Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102061167-102061432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC53
Entrez Gene ID
51019
Gene Name
coiled-coil domain containing 53
Gene Aliases
CGI-116
UniGene
Hs.405692
Gene Chromosome Location
Chr.12: 102012840-102062124 on Build GRCh38
Transcripts
NM_001301107.1 NM_016053.3 XM_011538423.1 XM_017019383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199996 c.109A>G p.T37A Chr.12: 102061289-102061289 on Build GRCh38
934080 c.55C>T p.P19S Chr.12: 102061343-102061343 on Build GRCh38
6462210 c.69G>A p.Q23Q Chr.12: 102061329-102061329 on Build GRCh38
5729895 c.143G>A p.C48Y Chr.12: 102061255-102061255 on Build GRCh38
934079 c.110C>T p.T37I Chr.12: 102061288-102061288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750922045 Chr.12: 102061240-102061240 on Build GRCh38
rs55932569 Chr.12: 102061325-102061325 on Build GRCh38
rs754601652 Chr.12: 102061348-102061348 on Build GRCh38
rs769052534 Chr.12: 102061295-102061295 on Build GRCh38
rs757855416 Chr.12: 102061244-102061244 on Build GRCh38
rs914758495 Chr.12: 102061245-102061245 on Build GRCh38
rs751109503 Chr.12: 102061335-102061335 on Build GRCh38
rs761037670 Chr.12: 102061218-102061218 on Build GRCh38
rs374994338 Chr.12: 102061236-102061236 on Build GRCh38
rs749197300 Chr.12: 102061286-102061286 on Build GRCh38
rs765521410 Chr.12: 102061306-102061306 on Build GRCh38
rs745639304 Chr.12: 102061387-102061387 on Build GRCh38
rs765721963 Chr.12: 102061239-102061239 on Build GRCh38
rs200289876 Chr.12: 102061266-102061266 on Build GRCh38
rs765884942 Chr.12: 102061247-102061247 on Build GRCh38
rs747401436 Chr.12: 102061250-102061250 on Build GRCh38
rs772181698 Chr.12: 102061287-102061287 on Build GRCh38
rs757304347 Chr.12: 102061394-102061394 on Build GRCh38
rs372954875 Chr.12: 102061269-102061269 on Build GRCh38
rs561071846 Chr.12: 102061389-102061389 on Build GRCh38
rs762194655 Chr.12: 102061305-102061305 on Build GRCh38
rs764552457 Chr.12: 102061228-102061228 on Build GRCh38
rs777179402 Chr.12: 102061230-102061230 on Build GRCh38
rs754691344 Chr.12: 102061263-102061263 on Build GRCh38
rs777700170 Chr.12: 102061273-102061273 on Build GRCh38
rs1047727028 Chr.12: 102061325-102061325 on Build GRCh38
rs777011712 Chr.12: 102061302-102061302 on Build GRCh38
rs765868302 Chr.12: 102061323-102061323 on Build GRCh38
rs201521376 Chr.12: 102061289-102061289 on Build GRCh38
rs767116456 Chr.12: 102061362-102061362 on Build GRCh38
rs377155225 Chr.12: 102061270-102061270 on Build GRCh38
rs768956856 Chr.12: 102061260-102061261 on Build GRCh38
rs145187756 Chr.12: 102061378-102061378 on Build GRCh38
rs763425493 Chr.12: 102061320-102061320 on Build GRCh38
rs755881197 Chr.12: 102061380-102061380 on Build GRCh38
rs907952665 Chr.12: 102061337-102061337 on Build GRCh38
rs751197817 Chr.12: 102061260-102061260 on Build GRCh38
rs752297365 Chr.12: 102061369-102061369 on Build GRCh38

View Full Product Details