Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205157198-205157463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTYK
Entrez Gene ID
25778
Gene Name
dual serine/threonine and tyrosine protein kinase
Gene Aliases
CAKUT1, DustyPK, HDCMD38P, RIP5, RIPK5
UniGene
Hs.6874
Gene Chromosome Location
Chr.1: 205142497-205211599 on Build GRCh38
Transcripts
NM_015375.2 NM_199462.2 XM_011509392.2 XM_011509393.2 XM_011509394.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400857 c.2352+3A>G p.? Chr.1: 205157270-205157270 on Build GRCh38
4806270 c.2244G>A p.G748G Chr.1: 205157381-205157381 on Build GRCh38
7261461 c.2302C>A p.L768M Chr.1: 205157323-205157323 on Build GRCh38
3803603 c.2341A>G p.K781E Chr.1: 205157284-205157284 on Build GRCh38
4027392 c.2290G>A p.G764R Chr.1: 205157335-205157335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184840693 Chr.1: 205157404-205157404 on Build GRCh38
rs773358240 Chr.1: 205157294-205157294 on Build GRCh38
rs372939615 Chr.1: 205157313-205157313 on Build GRCh38
rs879125937 Chr.1: 205157318-205157318 on Build GRCh38
rs760029067 Chr.1: 205157329-205157329 on Build GRCh38
rs768535879 Chr.1: 205157242-205157242 on Build GRCh38
rs142354055 Chr.1: 205157324-205157324 on Build GRCh38
rs34615388 Chr.1: 205157309-205157310 on Build GRCh38
rs377161801 Chr.1: 205157365-205157365 on Build GRCh38
rs955285572 Chr.1: 205157239-205157239 on Build GRCh38
rs776706190 Chr.1: 205157328-205157328 on Build GRCh38
rs770787598 Chr.1: 205157426-205157426 on Build GRCh38
rs558763686 Chr.1: 205157227-205157227 on Build GRCh38
rs991969050 Chr.1: 205157340-205157340 on Build GRCh38
rs749028852 Chr.1: 205157429-205157429 on Build GRCh38
rs935288326 Chr.1: 205157416-205157416 on Build GRCh38
rs756706640 Chr.1: 205157397-205157397 on Build GRCh38
rs111507314 Chr.1: 205157276-205157276 on Build GRCh38
rs769550887 Chr.1: 205157236-205157236 on Build GRCh38
rs371890151 Chr.1: 205157412-205157412 on Build GRCh38
rs745476293 Chr.1: 205157411-205157411 on Build GRCh38
rs753063101 Chr.1: 205157350-205157350 on Build GRCh38
rs978725505 Chr.1: 205157234-205157234 on Build GRCh38
rs137889690 Chr.1: 205157238-205157238 on Build GRCh38
rs764628186 Chr.1: 205157369-205157369 on Build GRCh38
rs770322382 Chr.1: 205157364-205157364 on Build GRCh38
rs749258992 Chr.1: 205157240-205157240 on Build GRCh38
rs765666210 Chr.1: 205157339-205157339 on Build GRCh38
rs776235566 Chr.1: 205157437-205157437 on Build GRCh38
rs34474155 Chr.1: 205157284-205157284 on Build GRCh38
rs780700974 Chr.1: 205157229-205157229 on Build GRCh38

View Full Product Details