Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93755171-93755444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf54
Entrez Gene ID
28970
Gene Name
chromosome 11 open reading frame 54
Gene Aliases
PTD012, PTOD012
UniGene
Hs.8360
Gene Chromosome Location
Chr.11: 93741594-93765323 on Build GRCh38
Transcripts
NM_001286067.1 NM_001286068.1 NM_001286069.1 NM_001286070.1 NM_001286071.1 NM_014039.3 XM_006718824.1 XM_011542781.2 XM_011542782.2 XM_017017613.1 XM_017017614.1 XM_017017615.1 XM_017017616.1 XM_017017617.1 XM_017017618.1 XM_017017619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891430 c.370C>T p.P124S Chr.11: 93755249-93755249 on Build GRCh38
5797225 c.429G>C p.E143D Chr.11: 93755308-93755308 on Build GRCh38
6135130 c.457C>T p.Q153* Chr.11: 93755336-93755336 on Build GRCh38
5797226 c.429G>C p.E143D Chr.11: 93755308-93755308 on Build GRCh38
5728925 c.352G>A p.E118K Chr.11: 93755231-93755231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754944862 Chr.11: 93755221-93755221 on Build GRCh38
rs201676030 Chr.11: 93755387-93755387 on Build GRCh38
rs867986510 Chr.11: 93755364-93755364 on Build GRCh38
rs777046889 Chr.11: 93755318-93755318 on Build GRCh38
rs760777056 Chr.11: 93755354-93755354 on Build GRCh38
rs965980904 Chr.11: 93755410-93755410 on Build GRCh38
rs759652495 Chr.11: 93755367-93755367 on Build GRCh38
rs773550494 Chr.11: 93755342-93755342 on Build GRCh38
rs778948483 Chr.11: 93755227-93755227 on Build GRCh38
rs928850857 Chr.11: 93755405-93755405 on Build GRCh38
rs528040589 Chr.11: 93755356-93755356 on Build GRCh38
rs774798574 Chr.11: 93755310-93755310 on Build GRCh38
rs544752502 Chr.11: 93755376-93755376 on Build GRCh38
rs769425143 Chr.11: 93755260-93755260 on Build GRCh38
rs780648367 Chr.11: 93755282-93755282 on Build GRCh38
rs990765563 Chr.11: 93755328-93755328 on Build GRCh38
rs756877146 Chr.11: 93755397-93755397 on Build GRCh38
rs755767885 Chr.11: 93755273-93755273 on Build GRCh38
rs752522273 Chr.11: 93755372-93755372 on Build GRCh38
rs747710006 Chr.11: 93755255-93755256 on Build GRCh38
rs200861861 Chr.11: 93755316-93755316 on Build GRCh38
rs765272344 Chr.11: 93755369-93755369 on Build GRCh38
rs201063419 Chr.11: 93755217-93755217 on Build GRCh38
rs749840078 Chr.11: 93755296-93755296 on Build GRCh38
rs994634968 Chr.11: 93755247-93755247 on Build GRCh38
rs145950304 Chr.11: 93755235-93755235 on Build GRCh38
rs772267032 Chr.11: 93755338-93755338 on Build GRCh38
rs376163608 Chr.11: 93755411-93755411 on Build GRCh38
rs138467282 Chr.11: 93755314-93755314 on Build GRCh38
rs371863678 Chr.11: 93755395-93755395 on Build GRCh38
rs758067298 Chr.11: 93755252-93755252 on Build GRCh38
rs753867854 Chr.11: 93755203-93755203 on Build GRCh38
rs371570110 Chr.11: 93755264-93755264 on Build GRCh38
rs1018855626 Chr.11: 93755403-93755403 on Build GRCh38
rs74351218 Chr.11: 93755315-93755315 on Build GRCh38
rs367758301 Chr.11: 93755393-93755393 on Build GRCh38
rs766562128 Chr.11: 93755202-93755202 on Build GRCh38
rs374717177 Chr.11: 93755306-93755306 on Build GRCh38
rs770490596 Chr.11: 93755281-93755281 on Build GRCh38
rs753995347 Chr.11: 93755361-93755361 on Build GRCh38

View Full Product Details