Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91891146-91891413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN5
Entrez Gene ID
10516
Gene Name
fibulin 5
Gene Aliases
ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, HNARMD, UP50
UniGene
Hs.332708
Gene Chromosome Location
Chr.14: 91869411-91947702 on Build GRCh38
Transcripts
NM_006329.3 XM_005267267.3 XM_011536356.1 XM_011536357.1 XM_011536358.1 XM_017020929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1371565 c.566C>A p.S189Y Chr.14: 91891274-91891274 on Build GRCh38
7133738 c.598G>A p.E200K Chr.14: 91891242-91891242 on Build GRCh38
3498950 c.590C>T p.T197I Chr.14: 91891250-91891250 on Build GRCh38
1678200 c.560C>T p.S187F Chr.14: 91891280-91891280 on Build GRCh38
6846908 c.596A>G p.N199S Chr.14: 91891244-91891244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941243257 Chr.14: 91891376-91891376 on Build GRCh38
rs534219640 Chr.14: 91891189-91891189 on Build GRCh38
rs752012733 Chr.14: 91891250-91891250 on Build GRCh38
rs767705850 Chr.14: 91891389-91891389 on Build GRCh38
rs769710145 Chr.14: 91891380-91891380 on Build GRCh38
rs753052522 Chr.14: 91891217-91891217 on Build GRCh38
rs147213753 Chr.14: 91891320-91891320 on Build GRCh38
rs1044103502 Chr.14: 91891392-91891392 on Build GRCh38
rs143767679 Chr.14: 91891294-91891294 on Build GRCh38
rs367710691 Chr.14: 91891200-91891200 on Build GRCh38
rs775878313 Chr.14: 91891359-91891359 on Build GRCh38
rs370854347 Chr.14: 91891211-91891211 on Build GRCh38
rs762167286 Chr.14: 91891388-91891388 on Build GRCh38
rs747934561 Chr.14: 91891290-91891290 on Build GRCh38
rs137901671 Chr.14: 91891235-91891235 on Build GRCh38
rs759056069 Chr.14: 91891207-91891207 on Build GRCh38
rs199912550 Chr.14: 91891286-91891286 on Build GRCh38
rs745867638 Chr.14: 91891244-91891244 on Build GRCh38
rs775352873 Chr.14: 91891387-91891387 on Build GRCh38
rs746858572 Chr.14: 91891305-91891305 on Build GRCh38
rs759297367 Chr.14: 91891336-91891336 on Build GRCh38
rs763330241 Chr.14: 91891371-91891371 on Build GRCh38
rs909797735 Chr.14: 91891335-91891335 on Build GRCh38
rs777892769 Chr.14: 91891296-91891296 on Build GRCh38
rs947347894 Chr.14: 91891364-91891364 on Build GRCh38
rs868328360 Chr.14: 91891300-91891300 on Build GRCh38
rs750670144 Chr.14: 91891259-91891259 on Build GRCh38
rs770310945 Chr.14: 91891192-91891192 on Build GRCh38
rs1055718757 Chr.14: 91891379-91891379 on Build GRCh38
rs1052990296 Chr.14: 91891295-91891295 on Build GRCh38
rs772898300 Chr.14: 91891174-91891174 on Build GRCh38
rs985149484 Chr.14: 91891196-91891196 on Build GRCh38
rs913136472 Chr.14: 91891352-91891352 on Build GRCh38
rs756288143 Chr.14: 91891267-91891267 on Build GRCh38
rs775989591 Chr.14: 91891332-91891332 on Build GRCh38
rs781064747 Chr.14: 91891256-91891256 on Build GRCh38
rs28939072 Chr.14: 91891334-91891334 on Build GRCh38
rs772104783 Chr.14: 91891171-91891171 on Build GRCh38
rs80338765 Chr.14: 91891236-91891236 on Build GRCh38
rs762077090 Chr.14: 91891376-91891376 on Build GRCh38
rs778893310 Chr.14: 91891275-91891275 on Build GRCh38
rs374908953 Chr.14: 91891353-91891353 on Build GRCh38
rs757388863 Chr.14: 91891251-91891251 on Build GRCh38

View Full Product Details