Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57301552-57301813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
UniGene
Hs.201378
Gene Chromosome Location
Chr.3: 57293699-57545203 on Build GRCh38
Transcripts
NM_001291661.1 XM_011533466.1 XM_011533467.1 XM_011533468.1 XM_011533469.1 XM_017005860.1 XM_017005861.1 XM_017005862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3824448 c.8778G>T p.L2926F Chr.3: 57301747-57301747 on Build GRCh38
3714601 c.8772C>A p.A2924A Chr.3: 57301753-57301753 on Build GRCh38
1738840 c.8780A>T p.N2927I Chr.3: 57301745-57301745 on Build GRCh38
1047633 c.8773C>T p.R2925W Chr.3: 57301752-57301752 on Build GRCh38
4616594 c.8784T>C p.F2928F Chr.3: 57301741-57301741 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774029444 Chr.3: 57301699-57301699 on Build GRCh38
rs765197537 Chr.3: 57301711-57301711 on Build GRCh38
rs768228450 Chr.3: 57301696-57301696 on Build GRCh38
rs757947796 Chr.3: 57301719-57301720 on Build GRCh38
rs200349291 Chr.3: 57301678-57301691 on Build GRCh38
rs757030626 Chr.3: 57301682-57301691 on Build GRCh38
rs62252001 Chr.3: 57301663-57301663 on Build GRCh38
rs116071948 Chr.3: 57301658-57301658 on Build GRCh38
rs113952788 Chr.3: 57301589-57301765 on Build GRCh38
rs3223090 Chr.3: 57301679-57301680 on Build GRCh38
rs956390342 Chr.3: 57301669-57301669 on Build GRCh38
rs1035932981 Chr.3: 57301717-57301727 on Build GRCh38
rs66545318 Chr.3: 57301715-57301728 on Build GRCh38
rs982717898 Chr.3: 57301792-57301792 on Build GRCh38
rs374403630 Chr.3: 57301770-57301770 on Build GRCh38
rs749000069 Chr.3: 57301688-57301688 on Build GRCh38
rs745418145 Chr.3: 57301715-57301727 on Build GRCh38
rs771563572 Chr.3: 57301715-57301715 on Build GRCh38
rs57748737 Chr.3: 57301719-57301728 on Build GRCh38
rs955496197 Chr.3: 57301764-57301764 on Build GRCh38
rs1012425303 Chr.3: 57301591-57301591 on Build GRCh38
rs922427166 Chr.3: 57301751-57301751 on Build GRCh38
rs878863136 Chr.3: 57301678-57301685 on Build GRCh38
rs67229457 Chr.3: 57301678-57301687 on Build GRCh38
rs761367228 Chr.3: 57301708-57301708 on Build GRCh38
rs878876985 Chr.3: 57301678-57301683 on Build GRCh38
rs747562232 Chr.3: 57301615-57301615 on Build GRCh38
rs760905563 Chr.3: 57301684-57301691 on Build GRCh38
rs984530249 Chr.3: 57301754-57301754 on Build GRCh38
rs1010954391 Chr.3: 57301707-57301707 on Build GRCh38
rs760559757 Chr.3: 57301741-57301741 on Build GRCh38
rs960274244 Chr.3: 57301725-57301725 on Build GRCh38
rs533042711 Chr.3: 57301785-57301785 on Build GRCh38
rs1052952678 Chr.3: 57301592-57301592 on Build GRCh38
rs71919228 Chr.3: 57301678-57301689 on Build GRCh38
rs184571375 Chr.3: 57301668-57301668 on Build GRCh38
rs1001470958 Chr.3: 57301669-57301672 on Build GRCh38
rs199748125 Chr.3: 57301771-57301771 on Build GRCh38
rs4681970 Chr.3: 57301719-57301719 on Build GRCh38
rs746981485 Chr.3: 57301726-57301729 on Build GRCh38
rs963740015 Chr.3: 57301727-57301727 on Build GRCh38

View Full Product Details