Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44973864-44974093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERB2
Entrez Gene ID
145645
Gene Name
telomere repeat binding bouquet formation protein 2
Gene Aliases
C15orf43
UniGene
Hs.567701
Gene Chromosome Location
Chr.15: 44956674-44979223 on Build GRCh38
Transcripts
NM_152448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5701943 c.487_488CC>GA p.P163D Chr.15: 44973919-44973920 on Build GRCh38
4666930 c.469G>A p.E157K Chr.15: 44973901-44973901 on Build GRCh38
5383390 c.517G>A p.V173I Chr.15: 44973949-44973949 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs564175883 Chr.15: 44973999-44973999 on Build GRCh38
rs765562252 Chr.15: 44973915-44973915 on Build GRCh38
rs978004457 Chr.15: 44974066-44974066 on Build GRCh38
rs745816415 Chr.15: 44974032-44974032 on Build GRCh38
rs34341793 Chr.15: 44973996-44973999 on Build GRCh38
rs918918375 Chr.15: 44974013-44974013 on Build GRCh38
rs757184485 Chr.15: 44973986-44973986 on Build GRCh38
rs865959196 Chr.15: 44973992-44973995 on Build GRCh38
rs201375705 Chr.15: 44973968-44973968 on Build GRCh38
rs747831131 Chr.15: 44973991-44973991 on Build GRCh38
rs990562763 Chr.15: 44973989-44973989 on Build GRCh38
rs985075804 Chr.15: 44974053-44974053 on Build GRCh38
rs541362165 Chr.15: 44973983-44973983 on Build GRCh38
rs755179803 Chr.15: 44973977-44973977 on Build GRCh38
rs913192144 Chr.15: 44974027-44974027 on Build GRCh38
rs751768068 Chr.15: 44973973-44973973 on Build GRCh38
rs772972851 Chr.15: 44973993-44973993 on Build GRCh38
rs758643789 Chr.15: 44973942-44973942 on Build GRCh38
rs1034393009 Chr.15: 44973971-44973971 on Build GRCh38
rs750831980 Chr.15: 44973921-44973921 on Build GRCh38
rs192772490 Chr.15: 44974050-44974050 on Build GRCh38
rs780440497 Chr.15: 44973972-44973972 on Build GRCh38
rs1001596889 Chr.15: 44973920-44973920 on Build GRCh38
rs577573803 Chr.15: 44974012-44974012 on Build GRCh38
rs781403498 Chr.15: 44973978-44973978 on Build GRCh38
rs970597674 Chr.15: 44973946-44973946 on Build GRCh38
rs904910337 Chr.15: 44973975-44973975 on Build GRCh38

View Full Product Details