Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232530088-232530315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRND
Entrez Gene ID
1144
Gene Name
cholinergic receptor nicotinic delta subunit
Gene Aliases
ACHRD, CMS2A, CMS3A, CMS3B, CMS3C, FCCMS, SCCMS
UniGene
Hs.156289
Gene Chromosome Location
Chr.2: 232526160-232537907 on Build GRCh38
Transcripts
NM_000751.2 NM_001256657.1 NM_001311195.1 NM_001311196.1 XM_011510524.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6835868 c.814G>A p.A272T Chr.2: 232530133-232530133 on Build GRCh38
4412373 c.820+9C>T p.? Chr.2: 232530148-232530148 on Build GRCh38
5736458 c.770G>A p.C257Y Chr.2: 232530089-232530089 on Build GRCh38
1017830 c.780C>T p.I260I Chr.2: 232530099-232530099 on Build GRCh38
3579106 c.777C>T p.L259L Chr.2: 232530096-232530096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560430528 Chr.2: 232530241-232530241 on Build GRCh38
rs775876937 Chr.2: 232530139-232530139 on Build GRCh38
rs1002190775 Chr.2: 232530114-232530114 on Build GRCh38
rs763880179 Chr.2: 232530152-232530152 on Build GRCh38
rs959978268 Chr.2: 232530162-232530162 on Build GRCh38
rs374813887 Chr.2: 232530179-232530179 on Build GRCh38
rs779733767 Chr.2: 232530164-232530164 on Build GRCh38
rs1055118152 Chr.2: 232530117-232530117 on Build GRCh38
rs760081896 Chr.2: 232530134-232530134 on Build GRCh38
rs769671837 Chr.2: 232530196-232530196 on Build GRCh38
rs532380193 Chr.2: 232530246-232530246 on Build GRCh38
rs920666922 Chr.2: 232530194-232530194 on Build GRCh38
rs372465644 Chr.2: 232530169-232530169 on Build GRCh38
rs1047843012 Chr.2: 232530201-232530201 on Build GRCh38
rs202209156 Chr.2: 232530136-232530136 on Build GRCh38
rs994961081 Chr.2: 232530125-232530125 on Build GRCh38
rs763425039 Chr.2: 232530190-232530190 on Build GRCh38
rs879255564 Chr.2: 232530139-232530140 on Build GRCh38
rs753625247 Chr.2: 232530168-232530168 on Build GRCh38
rs927227913 Chr.2: 232530257-232530257 on Build GRCh38
rs199984769 Chr.2: 232530187-232530187 on Build GRCh38
rs913084304 Chr.2: 232530197-232530197 on Build GRCh38
rs546219929 Chr.2: 232530277-232530277 on Build GRCh38
rs762545224 Chr.2: 232530146-232530146 on Build GRCh38
rs746384778 Chr.2: 232530184-232530184 on Build GRCh38
rs121909503 Chr.2: 232530131-232530131 on Build GRCh38
rs747127762 Chr.2: 232530115-232530115 on Build GRCh38
rs1045639164 Chr.2: 232530249-232530249 on Build GRCh38
rs751238043 Chr.2: 232530155-232530155 on Build GRCh38
rs750335170 Chr.2: 232530159-232530159 on Build GRCh38
rs540120869 Chr.2: 232530232-232530232 on Build GRCh38
rs944737292 Chr.2: 232530271-232530271 on Build GRCh38
rs771062973 Chr.2: 232530173-232530173 on Build GRCh38
rs368783371 Chr.2: 232530171-232530171 on Build GRCh38
rs745788897 Chr.2: 232530156-232530156 on Build GRCh38
rs147073853 Chr.2: 232530132-232530132 on Build GRCh38
rs141627135 Chr.2: 232530124-232530124 on Build GRCh38

View Full Product Details