Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 152108149-152108394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT11
Entrez Gene ID
63917
Gene Name
polypeptide N-acetylgalactosaminyltransferase 11
Gene Aliases
GALNAC-T11, GALNACT11
UniGene
Hs.647109
Gene Chromosome Location
Chr.7: 152025674-152122347 on Build GRCh38
Transcripts
NM_001304514.1 NM_022087.3 XM_006716082.2 XM_006716083.2 XM_006716084.2 XM_017012499.1 XM_017012500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781300 c.842C>T p.T281M Chr.7: 152108167-152108167 on Build GRCh38
4438857 c.857C>T p.S286L Chr.7: 152108182-152108182 on Build GRCh38
7204998 c.875G>A p.G292E Chr.7: 152108200-152108200 on Build GRCh38
143513 c.889_890GG>AA p.G297K Chr.7: 152108214-152108215 on Build GRCh38
4004381 c.935G>C p.R312P Chr.7: 152108260-152108260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758499061 Chr.7: 152108335-152108335 on Build GRCh38
rs769561206 Chr.7: 152108270-152108272 on Build GRCh38
rs748735399 Chr.7: 152108174-152108174 on Build GRCh38
rs755171248 Chr.7: 152108321-152108321 on Build GRCh38
rs35904688 Chr.7: 152108323-152108324 on Build GRCh38
rs141950078 Chr.7: 152108196-152108196 on Build GRCh38
rs764690207 Chr.7: 152108198-152108198 on Build GRCh38
rs535012637 Chr.7: 152108260-152108260 on Build GRCh38
rs772695869 Chr.7: 152108177-152108177 on Build GRCh38
rs774994840 Chr.7: 152108281-152108281 on Build GRCh38
rs749148717 Chr.7: 152108333-152108337 on Build GRCh38
rs777020469 Chr.7: 152108272-152108272 on Build GRCh38
rs202114668 Chr.7: 152108247-152108247 on Build GRCh38
rs764042419 Chr.7: 152108285-152108285 on Build GRCh38
rs201235145 Chr.7: 152108209-152108209 on Build GRCh38
rs773812713 Chr.7: 152108263-152108263 on Build GRCh38
rs533548373 Chr.7: 152108199-152108199 on Build GRCh38
rs1043744875 Chr.7: 152108271-152108271 on Build GRCh38
rs751501033 Chr.7: 152108295-152108295 on Build GRCh38
rs998575085 Chr.7: 152108342-152108342 on Build GRCh38
rs755227324 Chr.7: 152108256-152108256 on Build GRCh38
rs745749774 Chr.7: 152108189-152108189 on Build GRCh38
rs768410633 Chr.7: 152108333-152108333 on Build GRCh38
rs1031813009 Chr.7: 152108362-152108362 on Build GRCh38
rs767283433 Chr.7: 152108299-152108299 on Build GRCh38
rs757141823 Chr.7: 152108188-152108189 on Build GRCh38
rs769649251 Chr.7: 152108235-152108235 on Build GRCh38
rs371183866 Chr.7: 152108183-152108183 on Build GRCh38
rs1038646901 Chr.7: 152108239-152108239 on Build GRCh38
rs200681972 Chr.7: 152108264-152108264 on Build GRCh38
rs762381597 Chr.7: 152108283-152108283 on Build GRCh38
rs187517533 Chr.7: 152108259-152108259 on Build GRCh38
rs997011811 Chr.7: 152108316-152108316 on Build GRCh38
rs779214269 Chr.7: 152108330-152108330 on Build GRCh38
rs111668528 Chr.7: 152108334-152108334 on Build GRCh38
rs759176875 Chr.7: 152108276-152108276 on Build GRCh38
rs780864271 Chr.7: 152108169-152108169 on Build GRCh38
rs775484695 Chr.7: 152108286-152108289 on Build GRCh38
rs771247127 Chr.7: 152108266-152108266 on Build GRCh38
rs148150847 Chr.7: 152108168-152108168 on Build GRCh38
rs1056327561 Chr.7: 152108339-152108339 on Build GRCh38
rs752198002 Chr.7: 152108207-152108207 on Build GRCh38
rs751387828 Chr.7: 152108215-152108215 on Build GRCh38
rs745821568 Chr.7: 152108191-152108192 on Build GRCh38
rs371923465 Chr.7: 152108296-152108296 on Build GRCh38
rs766082693 Chr.7: 152108192-152108192 on Build GRCh38
rs750222167 Chr.7: 152108312-152108312 on Build GRCh38
rs993266562 Chr.7: 152108367-152108367 on Build GRCh38
rs756954040 Chr.7: 152108243-152108243 on Build GRCh38
rs896279279 Chr.7: 152108358-152108358 on Build GRCh38
rs749514801 Chr.7: 152108248-152108248 on Build GRCh38
rs981263195 Chr.7: 152108167-152108167 on Build GRCh38
rs776094150 Chr.7: 152108193-152108193 on Build GRCh38
rs570188437 Chr.7: 152108182-152108182 on Build GRCh38

View Full Product Details