Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41967587-41967860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A3
Entrez Gene ID
478
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Gene Aliases
AHC2, ATP1A1, CAPOS, DYT12, RDP
UniGene
Hs.515427
Gene Chromosome Location
Chr.19: 41966582-41994276 on Build GRCh38
Transcripts
NM_001256213.1 NM_001256214.1 NM_152296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078606 c.2842C>T p.L948L Chr.19: 41967741-41967741 on Build GRCh38
6084901 c.2853G>T p.E951D Chr.19: 41967730-41967730 on Build GRCh38
5056700 c.2895C>T p.D965D Chr.19: 41967688-41967688 on Build GRCh38
997165 c.2864C>A p.A955D Chr.19: 41967719-41967719 on Build GRCh38
3534545 c.2839G>A p.G947R Chr.19: 41967744-41967744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550881791 Chr.19: 41967610-41967610 on Build GRCh38
rs148292376 Chr.19: 41967727-41967727 on Build GRCh38
rs946947375 Chr.19: 41967739-41967739 on Build GRCh38
rs782411334 Chr.19: 41967760-41967760 on Build GRCh38
rs781954484 Chr.19: 41967803-41967803 on Build GRCh38
rs377619557 Chr.19: 41967623-41967623 on Build GRCh38
rs902680241 Chr.19: 41967788-41967788 on Build GRCh38
rs782303306 Chr.19: 41967691-41967691 on Build GRCh38
rs376361896 Chr.19: 41967648-41967648 on Build GRCh38
rs782422563 Chr.19: 41967650-41967650 on Build GRCh38
rs374318330 Chr.19: 41967635-41967635 on Build GRCh38
rs1040405243 Chr.19: 41967824-41967824 on Build GRCh38
rs606231446 Chr.19: 41967719-41967719 on Build GRCh38
rs200797477 Chr.19: 41967823-41967823 on Build GRCh38
rs369590943 Chr.19: 41967624-41967624 on Build GRCh38
rs781872061 Chr.19: 41967678-41967678 on Build GRCh38
rs886041431 Chr.19: 41967743-41967743 on Build GRCh38
rs567676147 Chr.19: 41967813-41967813 on Build GRCh38
rs782250260 Chr.19: 41967644-41967644 on Build GRCh38
rs782665893 Chr.19: 41967687-41967687 on Build GRCh38
rs398122887 Chr.19: 41967744-41967744 on Build GRCh38
rs190570469 Chr.19: 41967651-41967651 on Build GRCh38
rs769713426 Chr.19: 41967800-41967800 on Build GRCh38
rs782354767 Chr.19: 41967615-41967615 on Build GRCh38
rs141412861 Chr.19: 41967697-41967697 on Build GRCh38
rs782437931 Chr.19: 41967688-41967688 on Build GRCh38
rs534301907 Chr.19: 41967703-41967703 on Build GRCh38
rs1055403265 Chr.19: 41967723-41967723 on Build GRCh38
rs782611500 Chr.19: 41967676-41967676 on Build GRCh38
rs141421692 Chr.19: 41967745-41967745 on Build GRCh38
rs782159890 Chr.19: 41967632-41967632 on Build GRCh38
rs782381226 Chr.19: 41967712-41967712 on Build GRCh38
rs527543649 Chr.19: 41967818-41967818 on Build GRCh38
rs145179304 Chr.19: 41967698-41967698 on Build GRCh38
rs551838277 Chr.19: 41967679-41967679 on Build GRCh38
rs996107463 Chr.19: 41967834-41967834 on Build GRCh38
rs782133320 Chr.19: 41967806-41967806 on Build GRCh38
rs782766967 Chr.19: 41967798-41967798 on Build GRCh38
rs782757508 Chr.19: 41967628-41967628 on Build GRCh38
rs781900921 Chr.19: 41967629-41967629 on Build GRCh38
rs4306638 Chr.19: 41967831-41967831 on Build GRCh38
rs782204758 Chr.19: 41967706-41967706 on Build GRCh38
rs782754069 Chr.19: 41967809-41967809 on Build GRCh38
rs781941402 Chr.19: 41967785-41967785 on Build GRCh38

View Full Product Details