Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94533200-94533429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASD1
Entrez Gene ID
64921
Gene Name
CAS1 domain containing 1
Gene Aliases
C7orf12, NBLA04196
UniGene
Hs.260041
Gene Chromosome Location
Chr.7: 94509858-94633465 on Build GRCh38
Transcripts
NM_022900.4 XM_006716092.3 XM_006716093.3 XM_011516495.2 XM_017012546.1 XM_017012547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422924 c.484A>G p.I162V Chr.7: 94533229-94533229 on Build GRCh38
6411337 c.469G>A p.A157T Chr.7: 94533214-94533214 on Build GRCh38
1093276 c.488T>C p.V163A Chr.7: 94533233-94533233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376989588 Chr.7: 94533292-94533292 on Build GRCh38
rs369018635 Chr.7: 94533266-94533266 on Build GRCh38
rs904900828 Chr.7: 94533261-94533261 on Build GRCh38
rs202003082 Chr.7: 94533273-94533273 on Build GRCh38
rs994306407 Chr.7: 94533283-94533283 on Build GRCh38
rs765603845 Chr.7: 94533225-94533225 on Build GRCh38
rs914694883 Chr.7: 94533329-94533329 on Build GRCh38
rs947451296 Chr.7: 94533362-94533362 on Build GRCh38
rs780599033 Chr.7: 94533260-94533260 on Build GRCh38
rs984819120 Chr.7: 94533314-94533314 on Build GRCh38
rs374706874 Chr.7: 94533245-94533245 on Build GRCh38
rs772783074 Chr.7: 94533290-94533290 on Build GRCh38
rs371747135 Chr.7: 94533227-94533227 on Build GRCh38
rs373337938 Chr.7: 94533279-94533279 on Build GRCh38
rs1003278178 Chr.7: 94533262-94533262 on Build GRCh38
rs1011249107 Chr.7: 94533224-94533224 on Build GRCh38
rs541873074 Chr.7: 94533306-94533306 on Build GRCh38
rs537673362 Chr.7: 94533360-94533360 on Build GRCh38
rs977829001 Chr.7: 94533374-94533374 on Build GRCh38
rs576355522 Chr.7: 94533304-94533304 on Build GRCh38
rs555852751 Chr.7: 94533324-94533324 on Build GRCh38
rs922665777 Chr.7: 94533367-94533367 on Build GRCh38
rs911203031 Chr.7: 94533319-94533319 on Build GRCh38
rs747683332 Chr.7: 94533277-94533277 on Build GRCh38
rs774197143 Chr.7: 94533295-94533295 on Build GRCh38
rs750766469 Chr.7: 94533232-94533232 on Build GRCh38
rs1034798173 Chr.7: 94533272-94533272 on Build GRCh38
rs759503216 Chr.7: 94533301-94533301 on Build GRCh38
rs537385878 Chr.7: 94533271-94533271 on Build GRCh38
rs773864638 Chr.7: 94533363-94533363 on Build GRCh38
rs749093003 Chr.7: 94533291-94533291 on Build GRCh38

View Full Product Details