Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70429709-70429934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTH
Entrez Gene ID
1491
Gene Name
cystathionine gamma-lyase
Gene Aliases
-
UniGene
Hs.19904
Gene Chromosome Location
Chr.1: 70411218-70441949 on Build GRCh38
Transcripts
NM_001190463.1 NM_001902.5 NM_153742.4 XM_005270509.3 XM_017000416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426661 c.639C>G p.Y213* Chr.1: 70429844-70429844 on Build GRCh38
3985215 c.618T>C p.S206S Chr.1: 70429823-70429823 on Build GRCh38
5798144 c.589-7C>G p.? Chr.1: 70429787-70429787 on Build GRCh38
400794 c.589_590CG>TC p.R197>? Chr.1: 70429794-70429795 on Build GRCh38
5684386 c.640A>G p.M214V Chr.1: 70429845-70429845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038524981 Chr.1: 70429904-70429904 on Build GRCh38
rs745847430 Chr.1: 70429774-70429775 on Build GRCh38
rs377526667 Chr.1: 70429905-70429905 on Build GRCh38
rs751831301 Chr.1: 70429888-70429888 on Build GRCh38
rs764454576 Chr.1: 70429881-70429881 on Build GRCh38
rs1040477906 Chr.1: 70429779-70429779 on Build GRCh38
rs371610652 Chr.1: 70429787-70429787 on Build GRCh38
rs146873974 Chr.1: 70429824-70429824 on Build GRCh38
rs553130018 Chr.1: 70429868-70429868 on Build GRCh38
rs778867944 Chr.1: 70429745-70429745 on Build GRCh38
rs376661491 Chr.1: 70429825-70429825 on Build GRCh38
rs759962330 Chr.1: 70429856-70429856 on Build GRCh38
rs780110366 Chr.1: 70429774-70429774 on Build GRCh38
rs758722572 Chr.1: 70429760-70429760 on Build GRCh38
rs180856886 Chr.1: 70429827-70429827 on Build GRCh38
rs551720451 Chr.1: 70429794-70429794 on Build GRCh38
rs773402430 Chr.1: 70429763-70429763 on Build GRCh38
rs758347970 Chr.1: 70429785-70429785 on Build GRCh38
rs937494281 Chr.1: 70429898-70429898 on Build GRCh38
rs765732246 Chr.1: 70429858-70429858 on Build GRCh38
rs776993716 Chr.1: 70429849-70429849 on Build GRCh38
rs761095023 Chr.1: 70429845-70429845 on Build GRCh38
rs758172594 Chr.1: 70429744-70429744 on Build GRCh38
rs778096069 Chr.1: 70429762-70429762 on Build GRCh38
rs769684795 Chr.1: 70429764-70429764 on Build GRCh38
rs772819637 Chr.1: 70429836-70429836 on Build GRCh38
rs142475948 Chr.1: 70429795-70429795 on Build GRCh38
rs201568449 Chr.1: 70429791-70429791 on Build GRCh38
rs752753614 Chr.1: 70429748-70429748 on Build GRCh38
rs778067094 Chr.1: 70429861-70429861 on Build GRCh38

View Full Product Details