Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205528838-205529077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
UniGene
Hs.445402
Gene Chromosome Location
Chr.1: 205504556-205532793 on Build GRCh38
Transcripts
NM_002596.3 NM_212502.2 NM_212503.2 XM_011509602.2 XM_017001423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389042 c.1046A>G p.E349G Chr.1: 205529070-205529070 on Build GRCh38
6389043 c.1136A>G p.E379G Chr.1: 205529070-205529070 on Build GRCh38
6387037 c.1004C>T p.A335V Chr.1: 205529028-205529028 on Build GRCh38
6387038 c.1094C>T p.A365V Chr.1: 205529028-205529028 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886481882 Chr.1: 205528994-205528994 on Build GRCh38
rs772744995 Chr.1: 205529001-205529001 on Build GRCh38
rs937126377 Chr.1: 205528929-205528929 on Build GRCh38
rs554651789 Chr.1: 205528982-205528982 on Build GRCh38
rs774875965 Chr.1: 205528991-205528991 on Build GRCh38
rs749527541 Chr.1: 205528984-205528984 on Build GRCh38
rs927096670 Chr.1: 205528928-205528928 on Build GRCh38
rs746006314 Chr.1: 205528965-205528965 on Build GRCh38
rs920640814 Chr.1: 205528962-205528962 on Build GRCh38
rs534685414 Chr.1: 205528977-205528977 on Build GRCh38
rs974503565 Chr.1: 205528857-205528857 on Build GRCh38
rs768962329 Chr.1: 205528987-205528987 on Build GRCh38
rs200438485 Chr.1: 205529031-205529031 on Build GRCh38
rs981404475 Chr.1: 205528927-205528927 on Build GRCh38
rs932983993 Chr.1: 205528871-205528871 on Build GRCh38
rs7531230 Chr.1: 205528893-205528893 on Build GRCh38
rs558385581 Chr.1: 205528908-205528908 on Build GRCh38
rs373922103 Chr.1: 205528959-205528959 on Build GRCh38
rs900777096 Chr.1: 205529044-205529044 on Build GRCh38
rs957121310 Chr.1: 205528885-205528885 on Build GRCh38
rs370853296 Chr.1: 205528957-205528957 on Build GRCh38
rs764850479 Chr.1: 205529057-205529057 on Build GRCh38
rs989374941 Chr.1: 205528973-205528982 on Build GRCh38
rs1040658729 Chr.1: 205529038-205529038 on Build GRCh38
rs921709468 Chr.1: 205528866-205528866 on Build GRCh38
rs765924419 Chr.1: 205529014-205529014 on Build GRCh38
rs143048177 Chr.1: 205528917-205528917 on Build GRCh38
rs756309168 Chr.1: 205528959-205528960 on Build GRCh38
rs368089672 Chr.1: 205528963-205528963 on Build GRCh38
rs760156590 Chr.1: 205529003-205529003 on Build GRCh38
rs199542040 Chr.1: 205529059-205529059 on Build GRCh38
rs61733640 Chr.1: 205529027-205529027 on Build GRCh38

View Full Product Details