Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172665611-172665854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASLG
Entrez Gene ID
356
Gene Name
Fas ligand
Gene Aliases
ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6, TNLG1A
UniGene
Hs.2007
Gene Chromosome Location
Chr.1: 172659008-172666873 on Build GRCh38
Transcripts
NM_000639.2 NM_001302746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3934266 c.655C>T p.P219S Chr.1: 172665825-172665825 on Build GRCh38
3863555 c.541C>T p.L181F Chr.1: 172665711-172665711 on Build GRCh38
6459645 c.618C>A p.P206P Chr.1: 172665788-172665788 on Build GRCh38
3705246 c.655C>A p.P219T Chr.1: 172665825-172665825 on Build GRCh38
2075333 c.674T>C p.M225T Chr.1: 172665844-172665844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888867585 Chr.1: 172665644-172665644 on Build GRCh38
rs747293631 Chr.1: 172665715-172665715 on Build GRCh38
rs772212573 Chr.1: 172665702-172665702 on Build GRCh38
rs55728520 Chr.1: 172665819-172665819 on Build GRCh38
rs748115571 Chr.1: 172665695-172665695 on Build GRCh38
rs760032192 Chr.1: 172665732-172665732 on Build GRCh38
rs766268405 Chr.1: 172665807-172665807 on Build GRCh38
rs778663490 Chr.1: 172665832-172665832 on Build GRCh38
rs771262843 Chr.1: 172665722-172665722 on Build GRCh38
rs749218762 Chr.1: 172665663-172665663 on Build GRCh38
rs760432021 Chr.1: 172665667-172665667 on Build GRCh38
rs753616723 Chr.1: 172665809-172665809 on Build GRCh38
rs761180254 Chr.1: 172665794-172665794 on Build GRCh38
rs773263762 Chr.1: 172665711-172665711 on Build GRCh38
rs776079129 Chr.1: 172665759-172665759 on Build GRCh38
rs372784586 Chr.1: 172665692-172665692 on Build GRCh38
rs944346006 Chr.1: 172665710-172665710 on Build GRCh38
rs80358236 Chr.1: 172665642-172665725 on Build GRCh38
rs778993712 Chr.1: 172665685-172665685 on Build GRCh38
rs1043521093 Chr.1: 172665806-172665806 on Build GRCh38
rs147369993 Chr.1: 172665801-172665801 on Build GRCh38
rs559770662 Chr.1: 172665826-172665826 on Build GRCh38
rs12079514 Chr.1: 172665736-172665736 on Build GRCh38
rs374974805 Chr.1: 172665749-172665749 on Build GRCh38
rs541085313 Chr.1: 172665797-172665797 on Build GRCh38
rs900252191 Chr.1: 172665721-172665721 on Build GRCh38

View Full Product Details