Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80425791-80426064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213A
Entrez Gene ID
84293
Gene Name
family with sequence similarity 213 member A
Gene Aliases
C10orf58, PAMM
UniGene
Hs.500333
Gene Chromosome Location
Chr.10: 80407829-80437115 on Build GRCh38
Transcripts
NM_001243778.1 NM_001243779.1 NM_001243780.1 NM_001243781.1 NM_001243782.1 NM_032333.4 XM_011540265.2 XM_011540266.2 XM_017016775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6450758 c.310C>T p.Q104* Chr.10: 80425905-80425905 on Build GRCh38
685224 c.316G>T p.G106C Chr.10: 80425911-80425911 on Build GRCh38
685223 c.319G>A p.V107I Chr.10: 80425914-80425914 on Build GRCh38
920808 c.353G>T p.R118M Chr.10: 80425948-80425948 on Build GRCh38
3440995 c.399C>T p.F133F Chr.10: 80425994-80425994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951564389 Chr.10: 80425990-80425990 on Build GRCh38
rs953351672 Chr.10: 80425984-80425984 on Build GRCh38
rs577505930 Chr.10: 80425873-80425873 on Build GRCh38
rs757650152 Chr.10: 80425979-80425979 on Build GRCh38
rs751739734 Chr.10: 80425918-80425918 on Build GRCh38
rs201735051 Chr.10: 80426032-80426032 on Build GRCh38
rs781737207 Chr.10: 80425818-80425818 on Build GRCh38
rs200172480 Chr.10: 80425914-80425914 on Build GRCh38
rs777748216 Chr.10: 80425973-80425973 on Build GRCh38
rs767436522 Chr.10: 80425922-80425922 on Build GRCh38
rs769794297 Chr.10: 80425891-80425892 on Build GRCh38
rs762568858 Chr.10: 80426031-80426032 on Build GRCh38
rs749661744 Chr.10: 80425854-80425854 on Build GRCh38
rs776502753 Chr.10: 80426014-80426014 on Build GRCh38
rs763837127 Chr.10: 80426034-80426034 on Build GRCh38
rs746524695 Chr.10: 80426006-80426006 on Build GRCh38
rs548307191 Chr.10: 80425828-80425828 on Build GRCh38
rs141324021 Chr.10: 80425956-80425956 on Build GRCh38
rs755001031 Chr.10: 80425815-80425815 on Build GRCh38
rs376989414 Chr.10: 80425889-80425889 on Build GRCh38
rs768516415 Chr.10: 80426009-80426009 on Build GRCh38
rs534227355 Chr.10: 80425841-80425841 on Build GRCh38
rs755587236 Chr.10: 80425935-80425935 on Build GRCh38
rs776477369 Chr.10: 80425908-80425908 on Build GRCh38
rs760473330 Chr.10: 80425895-80425895 on Build GRCh38
rs143044329 Chr.10: 80425871-80425871 on Build GRCh38
rs189189429 Chr.10: 80425839-80425839 on Build GRCh38
rs774922306 Chr.10: 80425992-80425992 on Build GRCh38
rs368959911 Chr.10: 80426024-80426024 on Build GRCh38
rs771627730 Chr.10: 80425989-80425989 on Build GRCh38
rs200137753 Chr.10: 80425886-80425886 on Build GRCh38
rs141373617 Chr.10: 80425988-80425988 on Build GRCh38
rs778742142 Chr.10: 80425985-80425985 on Build GRCh38
rs1029121541 Chr.10: 80425970-80425970 on Build GRCh38
rs767771663 Chr.10: 80426025-80426025 on Build GRCh38
rs200341840 Chr.10: 80425849-80425849 on Build GRCh38
rs761573557 Chr.10: 80425913-80425913 on Build GRCh38
rs556273297 Chr.10: 80425926-80425926 on Build GRCh38
rs770751653 Chr.10: 80425865-80425865 on Build GRCh38
rs116495344 Chr.10: 80426033-80426033 on Build GRCh38
rs1017957166 Chr.10: 80425943-80425943 on Build GRCh38
rs774844546 Chr.10: 80425903-80425903 on Build GRCh38
rs546715508 Chr.10: 80425994-80425994 on Build GRCh38
rs764724246 Chr.10: 80426040-80426040 on Build GRCh38
rs372361112 Chr.10: 80425874-80425874 on Build GRCh38
rs761626656 Chr.10: 80426020-80426020 on Build GRCh38
rs148185355 Chr.10: 80425919-80425919 on Build GRCh38
rs192101938 Chr.10: 80425883-80425883 on Build GRCh38
rs749222156 Chr.10: 80426017-80426017 on Build GRCh38
rs764080064 Chr.10: 80425897-80425897 on Build GRCh38
rs764944371 Chr.10: 80426023-80426023 on Build GRCh38

View Full Product Details