Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89645843-89646116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP1A
Entrez Gene ID
5119
Gene Name
charged multivesicular body protein 1A
Gene Aliases
CHMP1, PCH8, PCOLN3, PRSM1, VPS46-1, VPS46A
UniGene
Hs.589427
Gene Chromosome Location
Chr.16: 89644431-89657785 on Build GRCh38
Transcripts
NM_001083314.3 NM_002768.4 NR_046418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5782532 c.628C>G p.P210A Chr.16: 89646009-89646009 on Build GRCh38
974915 c.641C>T p.A214V Chr.16: 89645996-89645996 on Build GRCh38
6628461 c.639C>T p.T213T Chr.16: 89645998-89645998 on Build GRCh38
119930 c.679G>C p.A227P Chr.16: 89645958-89645958 on Build GRCh38
6628462 c.635C>G p.P212R Chr.16: 89646002-89646002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747262339 Chr.16: 89645940-89645940 on Build GRCh38
rs756688699 Chr.16: 89645904-89645904 on Build GRCh38
rs752943661 Chr.16: 89646044-89646044 on Build GRCh38
rs370304473 Chr.16: 89645981-89645981 on Build GRCh38
rs764970236 Chr.16: 89645984-89645984 on Build GRCh38
rs201865392 Chr.16: 89646054-89646054 on Build GRCh38
rs61747591 Chr.16: 89645971-89645971 on Build GRCh38
rs754982493 Chr.16: 89646041-89646041 on Build GRCh38
rs778195826 Chr.16: 89646047-89646047 on Build GRCh38
rs201259282 Chr.16: 89646032-89646032 on Build GRCh38
rs374723745 Chr.16: 89645939-89645939 on Build GRCh38
rs765779716 Chr.16: 89645879-89645879 on Build GRCh38
rs760403388 Chr.16: 89645976-89645976 on Build GRCh38
rs777737345 Chr.16: 89645897-89645897 on Build GRCh38
rs751510291 Chr.16: 89646039-89646039 on Build GRCh38
rs369693521 Chr.16: 89645886-89645886 on Build GRCh38
rs116954456 Chr.16: 89645972-89645972 on Build GRCh38
rs116776046 Chr.16: 89645945-89645945 on Build GRCh38
rs775131172 Chr.16: 89646000-89646000 on Build GRCh38
rs755936611 Chr.16: 89645892-89645892 on Build GRCh38
rs771412753 Chr.16: 89645999-89645999 on Build GRCh38
rs773045170 Chr.16: 89646070-89646070 on Build GRCh38
rs375366854 Chr.16: 89645957-89645957 on Build GRCh38
rs756331359 Chr.16: 89646045-89646045 on Build GRCh38
rs117555004 Chr.16: 89645966-89645966 on Build GRCh38
rs367970292 Chr.16: 89645998-89645998 on Build GRCh38
rs746019338 Chr.16: 89645924-89645924 on Build GRCh38
rs746462901 Chr.16: 89646007-89646007 on Build GRCh38
rs767256166 Chr.16: 89645889-89645889 on Build GRCh38
rs775180423 Chr.16: 89645974-89645974 on Build GRCh38
rs747801608 Chr.16: 89646063-89646063 on Build GRCh38
rs900093048 Chr.16: 89645949-89645949 on Build GRCh38
rs751704834 Chr.16: 89645995-89645995 on Build GRCh38
rs373295230 Chr.16: 89645954-89645954 on Build GRCh38
rs763955537 Chr.16: 89645959-89645959 on Build GRCh38
rs888647550 Chr.16: 89645927-89645927 on Build GRCh38
rs753808559 Chr.16: 89645916-89645916 on Build GRCh38
rs567580372 Chr.16: 89645890-89645890 on Build GRCh38
rs767608796 Chr.16: 89646042-89646042 on Build GRCh38
rs772060654 Chr.16: 89645928-89645928 on Build GRCh38
rs763900642 Chr.16: 89645980-89645980 on Build GRCh38
rs372222150 Chr.16: 89646064-89646064 on Build GRCh38
rs759022850 Chr.16: 89645950-89645950 on Build GRCh38
rs766414872 Chr.16: 89645994-89645994 on Build GRCh38
rs776380381 Chr.16: 89646015-89646015 on Build GRCh38
rs768229451 Chr.16: 89646059-89646059 on Build GRCh38
rs372255335 Chr.16: 89645921-89645921 on Build GRCh38
rs781332646 Chr.16: 89645997-89645997 on Build GRCh38
rs762596072 Chr.16: 89646091-89646091 on Build GRCh38
rs757300622 Chr.16: 89645918-89645918 on Build GRCh38
rs757693176 Chr.16: 89645983-89645983 on Build GRCh38
rs913060528 Chr.16: 89645978-89645978 on Build GRCh38
rs542664777 Chr.16: 89645961-89645961 on Build GRCh38
rs761392378 Chr.16: 89646029-89646029 on Build GRCh38
rs766361313 Chr.16: 89646035-89646035 on Build GRCh38
rs750322473 Chr.16: 89645987-89645987 on Build GRCh38
rs750329191 Chr.16: 89645962-89645962 on Build GRCh38
rs768288651 Chr.16: 89646009-89646009 on Build GRCh38
rs759645131 Chr.16: 89646040-89646041 on Build GRCh38
rs746411478 Chr.16: 89646057-89646057 on Build GRCh38
rs921735430 Chr.16: 89645869-89645869 on Build GRCh38
rs373074379 Chr.16: 89645963-89645963 on Build GRCh38
rs542215121 Chr.16: 89645973-89645973 on Build GRCh38
rs773717647 Chr.16: 89645947-89645947 on Build GRCh38
rs776051113 Chr.16: 89646060-89646060 on Build GRCh38
rs758312752 Chr.16: 89645993-89645993 on Build GRCh38
rs369507937 Chr.16: 89645960-89645960 on Build GRCh38
rs1057108932 Chr.16: 89646080-89646080 on Build GRCh38
rs762943349 Chr.16: 89646033-89646033 on Build GRCh38
rs762330242 Chr.16: 89645872-89645872 on Build GRCh38
rs749676630 Chr.16: 89646053-89646053 on Build GRCh38
rs780126387 Chr.16: 89645964-89645964 on Build GRCh38
rs374390578 Chr.16: 89645996-89645996 on Build GRCh38
rs368448623 Chr.16: 89646056-89646056 on Build GRCh38
rs200889805 Chr.16: 89645958-89645958 on Build GRCh38
rs754579486 Chr.16: 89645887-89645887 on Build GRCh38

View Full Product Details