Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5748109-5748382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
UniGene
Hs.646899
Gene Chromosome Location
Chr.4: 5711197-5829043 on Build GRCh38
Transcripts
NM_001306090.1 NM_001306092.1 NM_153717.2 XM_006713865.3 XM_006713866.3 XM_011513419.2 XM_017007883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3775910 c.1054G>T p.A352S Chr.4: 5748262-5748262 on Build GRCh38
6511987 c.1097T>A p.L366Q Chr.4: 5748305-5748305 on Build GRCh38
179284 c.1074C>T p.C358C Chr.4: 5748282-5748282 on Build GRCh38
355201 c.1041G>T p.T347T Chr.4: 5748249-5748249 on Build GRCh38
6100779 c.1093G>T p.A365S Chr.4: 5748301-5748301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199916502 Chr.4: 5748190-5748190 on Build GRCh38
rs769839092 Chr.4: 5748193-5748193 on Build GRCh38
rs777624936 Chr.4: 5748143-5748143 on Build GRCh38
rs755931960 Chr.4: 5748324-5748324 on Build GRCh38
rs745627738 Chr.4: 5748255-5748256 on Build GRCh38
rs763807443 Chr.4: 5748235-5748235 on Build GRCh38
rs4688963 Chr.4: 5748177-5748177 on Build GRCh38
rs756025432 Chr.4: 5748267-5748267 on Build GRCh38
rs769651463 Chr.4: 5748178-5748178 on Build GRCh38
rs201738879 Chr.4: 5748133-5748133 on Build GRCh38
rs780972401 Chr.4: 5748170-5748170 on Build GRCh38
rs150923920 Chr.4: 5748283-5748283 on Build GRCh38
rs778893728 Chr.4: 5748334-5748334 on Build GRCh38
rs774029262 Chr.4: 5748318-5748318 on Build GRCh38
rs762140246 Chr.4: 5748209-5748209 on Build GRCh38
rs773476300 Chr.4: 5748225-5748225 on Build GRCh38
rs921573510 Chr.4: 5748282-5748282 on Build GRCh38
rs201090942 Chr.4: 5748338-5748338 on Build GRCh38
rs6854138 Chr.4: 5748304-5748304 on Build GRCh38
rs148418233 Chr.4: 5748249-5748249 on Build GRCh38
rs912969560 Chr.4: 5748310-5748310 on Build GRCh38
rs200707006 Chr.4: 5748136-5748136 on Build GRCh38
rs557010916 Chr.4: 5748148-5748148 on Build GRCh38
rs763650726 Chr.4: 5748311-5748311 on Build GRCh38
rs771462565 Chr.4: 5748302-5748302 on Build GRCh38
rs774625199 Chr.4: 5748205-5748205 on Build GRCh38
rs761463466 Chr.4: 5748319-5748319 on Build GRCh38
rs771137452 Chr.4: 5748205-5748206 on Build GRCh38
rs764011269 Chr.4: 5748323-5748323 on Build GRCh38
rs377390727 Chr.4: 5748223-5748223 on Build GRCh38
rs775052481 Chr.4: 5748227-5748227 on Build GRCh38
rs755096874 Chr.4: 5748272-5748272 on Build GRCh38
rs563257565 Chr.4: 5748321-5748321 on Build GRCh38
rs750190123 Chr.4: 5748263-5748263 on Build GRCh38
rs374621067 Chr.4: 5748134-5748134 on Build GRCh38
rs33929747 Chr.4: 5748276-5748276 on Build GRCh38
rs886043999 Chr.4: 5748290-5748290 on Build GRCh38
rs762531567 Chr.4: 5748306-5748306 on Build GRCh38
rs370388849 Chr.4: 5748301-5748301 on Build GRCh38
rs766312086 Chr.4: 5748326-5748326 on Build GRCh38
rs767093289 Chr.4: 5748320-5748320 on Build GRCh38
rs772431066 Chr.4: 5748211-5748211 on Build GRCh38
rs780118761 Chr.4: 5748189-5748189 on Build GRCh38
rs753723092 Chr.4: 5748328-5748328 on Build GRCh38
rs767913372 Chr.4: 5748242-5748243 on Build GRCh38
rs895165309 Chr.4: 5748213-5748213 on Build GRCh38
rs34947207 Chr.4: 5748248-5748248 on Build GRCh38
rs749331734 Chr.4: 5748152-5748152 on Build GRCh38
rs878989215 Chr.4: 5748317-5748317 on Build GRCh38
rs4688962 Chr.4: 5748234-5748234 on Build GRCh38
rs201991931 Chr.4: 5748347-5748347 on Build GRCh38
rs952552516 Chr.4: 5748342-5748342 on Build GRCh38
rs772185282 Chr.4: 5748291-5748291 on Build GRCh38
rs777924778 Chr.4: 5748349-5748349 on Build GRCh38
rs779915989 Chr.4: 5748268-5748268 on Build GRCh38
rs4688961 Chr.4: 5748357-5748357 on Build GRCh38
rs1057517875 Chr.4: 5748244-5748245 on Build GRCh38
rs773566770 Chr.4: 5748297-5748297 on Build GRCh38
rs768854884 Chr.4: 5748196-5748196 on Build GRCh38
rs754937564 Chr.4: 5748333-5748333 on Build GRCh38
rs121908425 Chr.4: 5748226-5748226 on Build GRCh38
rs757159611 Chr.4: 5748150-5748150 on Build GRCh38
rs375181612 Chr.4: 5748343-5748343 on Build GRCh38

View Full Product Details