Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14521119-14521390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107B
Entrez Gene ID
83641
Gene Name
family with sequence similarity 107 member B
Gene Aliases
C10orf45
UniGene
Hs.446315
Gene Chromosome Location
Chr.10: 14518557-14774897 on Build GRCh38
Transcripts
NM_001282695.1 NM_001282696.1 NM_001282697.1 NM_001282698.1 NM_001282699.1 NM_001282700.1 NM_001282701.1 NM_001282702.1 NM_001282703.1 NM_001320735.1 NM_001320736.1 NM_001320737.1 NM_001320738.1 NM_001320739.1 NM_001320740.1 NM_001320741.1 NM_031453.3 XM_005252616.3 XM_011519735.1 XM_017016746.1 XM_017016747.1 XM_017016748.1 XM_017016749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936558 c.913G>T p.E305* Chr.10: 14521198-14521198 on Build GRCh38
5468691 c.824A>T p.K275I Chr.10: 14521287-14521287 on Build GRCh38
3806830 c.873C>T p.G291G Chr.10: 14521238-14521238 on Build GRCh38
4750240 c.844A>C p.N282H Chr.10: 14521267-14521267 on Build GRCh38
3935002 c.830A>C p.Q277P Chr.10: 14521281-14521281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762175059 Chr.10: 14521204-14521204 on Build GRCh38
rs553992860 Chr.10: 14521170-14521170 on Build GRCh38
rs374866652 Chr.10: 14521298-14521298 on Build GRCh38
rs754480262 Chr.10: 14521329-14521329 on Build GRCh38
rs755622194 Chr.10: 14521160-14521160 on Build GRCh38
rs759715125 Chr.10: 14521176-14521176 on Build GRCh38
rs373444074 Chr.10: 14521272-14521272 on Build GRCh38
rs764141565 Chr.10: 14521201-14521201 on Build GRCh38
rs773495389 Chr.10: 14521168-14521168 on Build GRCh38
rs902221723 Chr.10: 14521245-14521245 on Build GRCh38
rs202167032 Chr.10: 14521248-14521248 on Build GRCh38
rs556308750 Chr.10: 14521276-14521276 on Build GRCh38
rs765312731 Chr.10: 14521178-14521178 on Build GRCh38
rs200526867 Chr.10: 14521312-14521312 on Build GRCh38
rs779476069 Chr.10: 14521162-14521162 on Build GRCh38
rs187116118 Chr.10: 14521210-14521210 on Build GRCh38
rs994635270 Chr.10: 14521274-14521274 on Build GRCh38
rs761894001 Chr.10: 14521299-14521299 on Build GRCh38
rs755722311 Chr.10: 14521306-14521306 on Build GRCh38
rs754493541 Chr.10: 14521275-14521277 on Build GRCh38
rs747581664 Chr.10: 14521262-14521262 on Build GRCh38
rs760651219 Chr.10: 14521352-14521352 on Build GRCh38
rs79621927 Chr.10: 14521167-14521167 on Build GRCh38
rs375536594 Chr.10: 14521224-14521224 on Build GRCh38
rs771252967 Chr.10: 14521263-14521263 on Build GRCh38
rs777027306 Chr.10: 14521264-14521264 on Build GRCh38
rs372209526 Chr.10: 14521223-14521223 on Build GRCh38
rs753584431 Chr.10: 14521315-14521315 on Build GRCh38
rs771342568 Chr.10: 14521173-14521173 on Build GRCh38
rs762933528 Chr.10: 14521282-14521282 on Build GRCh38
rs545321031 Chr.10: 14521301-14521301 on Build GRCh38
rs750982759 Chr.10: 14521351-14521351 on Build GRCh38
rs758878466 Chr.10: 14521256-14521256 on Build GRCh38
rs780982296 Chr.10: 14521152-14521152 on Build GRCh38
rs191623973 Chr.10: 14521265-14521265 on Build GRCh38
rs767464134 Chr.10: 14521216-14521216 on Build GRCh38
rs745586649 Chr.10: 14521159-14521159 on Build GRCh38
rs34265970 Chr.10: 14521300-14521300 on Build GRCh38
rs775464555 Chr.10: 14521180-14521180 on Build GRCh38
rs778239757 Chr.10: 14521259-14521259 on Build GRCh38
rs762882314 Chr.10: 14521187-14521187 on Build GRCh38
rs779573515 Chr.10: 14521236-14521236 on Build GRCh38
rs748940762 Chr.10: 14521227-14521229 on Build GRCh38
rs776828253 Chr.10: 14521174-14521174 on Build GRCh38
rs367780011 Chr.10: 14521211-14521211 on Build GRCh38
rs764339947 Chr.10: 14521283-14521283 on Build GRCh38
rs778380584 Chr.10: 14521309-14521309 on Build GRCh38

View Full Product Details