Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33623166-33623391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBFA2T2
Entrez Gene ID
9139
Gene Name
CBFA2/RUNX1 translocation partner 2
Gene Aliases
EHT, MTGR1, ZMYND3, p85
UniGene
Hs.153934
Gene Chromosome Location
Chr.20: 33490068-33650031 on Build GRCh38
Transcripts
NM_001032999.2 NM_001039709.1 NM_005093.3 XM_011529101.2 XM_011529102.2 XM_011529103.2 XM_011529107.2 XM_017028121.1 XM_017028122.1 XM_017028123.1 XM_017028124.1 XM_017028125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6287880 c.631C>T p.L211L Chr.20: 33623208-33623208 on Build GRCh38
1025954 c.603C>T p.S201S Chr.20: 33623180-33623180 on Build GRCh38
6299643 c.684G>A p.E228E Chr.20: 33623261-33623261 on Build GRCh38
7416596 c.670G>A p.G224R Chr.20: 33623274-33623274 on Build GRCh38
7152277 c.636C>G p.L212L Chr.20: 33623213-33623213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977767830 Chr.20: 33623253-33623253 on Build GRCh38
rs529484931 Chr.20: 33623273-33623273 on Build GRCh38
rs201708482 Chr.20: 33623199-33623199 on Build GRCh38
rs769979005 Chr.20: 33623324-33623324 on Build GRCh38
rs17124834 Chr.20: 33623337-33623337 on Build GRCh38
rs143592356 Chr.20: 33623357-33623357 on Build GRCh38
rs764101572 Chr.20: 33623247-33623247 on Build GRCh38
rs139409587 Chr.20: 33623198-33623198 on Build GRCh38
rs374071030 Chr.20: 33623272-33623272 on Build GRCh38
rs143525698 Chr.20: 33623264-33623264 on Build GRCh38
rs368244146 Chr.20: 33623328-33623328 on Build GRCh38
rs771290433 Chr.20: 33623189-33623189 on Build GRCh38
rs544838236 Chr.20: 33623294-33623294 on Build GRCh38
rs147158236 Chr.20: 33623267-33623267 on Build GRCh38
rs750301541 Chr.20: 33623268-33623268 on Build GRCh38
rs370737889 Chr.20: 33623190-33623190 on Build GRCh38
rs771359510 Chr.20: 33623307-33623307 on Build GRCh38
rs115034117 Chr.20: 33623362-33623362 on Build GRCh38
rs201083985 Chr.20: 33623319-33623319 on Build GRCh38
rs776972051 Chr.20: 33623317-33623317 on Build GRCh38
rs778320078 Chr.20: 33623300-33623300 on Build GRCh38
rs551482570 Chr.20: 33623336-33623336 on Build GRCh38
rs751472025 Chr.20: 33623257-33623257 on Build GRCh38
rs985214984 Chr.20: 33623287-33623287 on Build GRCh38
rs769157020 Chr.20: 33623205-33623207 on Build GRCh38
rs747487263 Chr.20: 33623301-33623301 on Build GRCh38
rs967384701 Chr.20: 33623231-33623231 on Build GRCh38
rs1008063428 Chr.20: 33623339-33623339 on Build GRCh38
rs752739929 Chr.20: 33623243-33623243 on Build GRCh38
rs763065839 Chr.20: 33623246-33623246 on Build GRCh38
rs765506102 Chr.20: 33623219-33623219 on Build GRCh38
rs372314464 Chr.20: 33623348-33623348 on Build GRCh38
rs761112426 Chr.20: 33623186-33623186 on Build GRCh38
rs772398282 Chr.20: 33623310-33623310 on Build GRCh38
rs941162333 Chr.20: 33623318-33623318 on Build GRCh38
rs527768632 Chr.20: 33623354-33623354 on Build GRCh38
rs748786648 Chr.20: 33623276-33623276 on Build GRCh38

View Full Product Details