Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2539440-2539671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
UniGene
Hs.520623
Gene Chromosome Location
Chr.7: 2537810-2555758 on Build GRCh38
Transcripts
NM_152743.3 XM_005249643.4 XM_011515177.2 XM_011515178.1 XM_011515179.2 XM_011515181.2 XM_011515182.2 XM_011515183.1 XM_011515184.2 XM_011515186.2 XM_017011833.1 XM_017011834.1 XM_017011835.1 XM_017011836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366898 c.1507C>T p.P503S Chr.7: 2539634-2539634 on Build GRCh38
6612481 c.1539C>T p.C513C Chr.7: 2539602-2539602 on Build GRCh38
6361777 c.1533C>A p.H511Q Chr.7: 2539608-2539608 on Build GRCh38
5645514 c.1558G>A p.A520T Chr.7: 2539583-2539583 on Build GRCh38
6428669 c.1538G>A p.C513Y Chr.7: 2539603-2539603 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760559426 Chr.7: 2539497-2539498 on Build GRCh38
rs758539173 Chr.7: 2539518-2539518 on Build GRCh38
rs370879134 Chr.7: 2539650-2539650 on Build GRCh38
rs112755163 Chr.7: 2539522-2539522 on Build GRCh38
rs777173605 Chr.7: 2539587-2539587 on Build GRCh38
rs902079997 Chr.7: 2539488-2539488 on Build GRCh38
rs771294370 Chr.7: 2539584-2539584 on Build GRCh38
rs779927413 Chr.7: 2539545-2539545 on Build GRCh38
rs533358915 Chr.7: 2539520-2539520 on Build GRCh38
rs763100774 Chr.7: 2539614-2539614 on Build GRCh38
rs754111516 Chr.7: 2539505-2539505 on Build GRCh38
rs747583240 Chr.7: 2539578-2539578 on Build GRCh38
rs761031060 Chr.7: 2539503-2539503 on Build GRCh38
rs145509776 Chr.7: 2539577-2539577 on Build GRCh38
rs142129866 Chr.7: 2539547-2539547 on Build GRCh38
rs768471054 Chr.7: 2539557-2539557 on Build GRCh38
rs151161353 Chr.7: 2539562-2539562 on Build GRCh38
rs766270301 Chr.7: 2539624-2539624 on Build GRCh38
rs751548415 Chr.7: 2539619-2539619 on Build GRCh38
rs755075934 Chr.7: 2539551-2539551 on Build GRCh38
rs147745609 Chr.7: 2539634-2539634 on Build GRCh38
rs765579812 Chr.7: 2539600-2539600 on Build GRCh38
rs754789952 Chr.7: 2539635-2539635 on Build GRCh38
rs754345213 Chr.7: 2539505-2539505 on Build GRCh38
rs759858132 Chr.7: 2539591-2539591 on Build GRCh38
rs755934097 Chr.7: 2539527-2539527 on Build GRCh38
rs753205052 Chr.7: 2539633-2539633 on Build GRCh38
rs200635453 Chr.7: 2539508-2539508 on Build GRCh38
rs140095103 Chr.7: 2539571-2539571 on Build GRCh38
rs535969254 Chr.7: 2539623-2539623 on Build GRCh38
rs376165927 Chr.7: 2539613-2539613 on Build GRCh38
rs114267478 Chr.7: 2539500-2539500 on Build GRCh38
rs373611568 Chr.7: 2539509-2539509 on Build GRCh38
rs778795414 Chr.7: 2539649-2539649 on Build GRCh38
rs963293277 Chr.7: 2539498-2539498 on Build GRCh38
rs377427021 Chr.7: 2539618-2539618 on Build GRCh38
rs771707994 Chr.7: 2539499-2539499 on Build GRCh38
rs370271691 Chr.7: 2539523-2539523 on Build GRCh38
rs1037045280 Chr.7: 2539468-2539468 on Build GRCh38

View Full Product Details