Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34868849-34869076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP3
Entrez Gene ID
58512
Gene Name
DLG associated protein 3
Gene Aliases
DAP3, SAPAP3
UniGene
Hs.733945
Gene Chromosome Location
Chr.1: 34862120-34929585 on Build GRCh38
Transcripts
NM_001080418.2 XM_011541879.2 XM_011541880.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4789380 c.2119C>A p.R707S Chr.1: 34868971-34868971 on Build GRCh38
6856499 c.2169delC p.T724fs*52 Chr.1: 34868921-34868921 on Build GRCh38
2079930 c.2017C>T p.R673C Chr.1: 34869073-34869073 on Build GRCh38
6997639 c.2189C>T p.T730M Chr.1: 34868901-34868901 on Build GRCh38
3488841 c.2106C>T p.A702A Chr.1: 34868984-34868984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370493091 Chr.1: 34868957-34868957 on Build GRCh38
rs768833299 Chr.1: 34868912-34868912 on Build GRCh38
rs542950831 Chr.1: 34868870-34868870 on Build GRCh38
rs776832849 Chr.1: 34868997-34868997 on Build GRCh38
rs771067661 Chr.1: 34868891-34868891 on Build GRCh38
rs772709051 Chr.1: 34868928-34868928 on Build GRCh38
rs754834169 Chr.1: 34868935-34868935 on Build GRCh38
rs756086794 Chr.1: 34868952-34868952 on Build GRCh38
rs367604760 Chr.1: 34869006-34869006 on Build GRCh38
rs779200016 Chr.1: 34868954-34868954 on Build GRCh38
rs774076646 Chr.1: 34869050-34869050 on Build GRCh38
rs762454988 Chr.1: 34869011-34869011 on Build GRCh38
rs772418605 Chr.1: 34868904-34868904 on Build GRCh38
rs759090974 Chr.1: 34869052-34869052 on Build GRCh38
rs150619481 Chr.1: 34868900-34868900 on Build GRCh38
rs747997191 Chr.1: 34868881-34868881 on Build GRCh38
rs373723836 Chr.1: 34868971-34868971 on Build GRCh38
rs758490483 Chr.1: 34868970-34868970 on Build GRCh38
rs765186393 Chr.1: 34868965-34868966 on Build GRCh38
rs766034075 Chr.1: 34868930-34868930 on Build GRCh38
rs565640559 Chr.1: 34869047-34869047 on Build GRCh38
rs770275838 Chr.1: 34869008-34869008 on Build GRCh38
rs769084336 Chr.1: 34868983-34868983 on Build GRCh38
rs367984704 Chr.1: 34868939-34868939 on Build GRCh38
rs746132929 Chr.1: 34868901-34868901 on Build GRCh38
rs767170740 Chr.1: 34868936-34868936 on Build GRCh38
rs866905158 Chr.1: 34868976-34868976 on Build GRCh38
rs199537135 Chr.1: 34868967-34868967 on Build GRCh38
rs765835465 Chr.1: 34869020-34869020 on Build GRCh38
rs773655239 Chr.1: 34869009-34869009 on Build GRCh38
rs781194450 Chr.1: 34868871-34868871 on Build GRCh38
rs764770295 Chr.1: 34868924-34868924 on Build GRCh38
rs747410186 Chr.1: 34868911-34868911 on Build GRCh38
rs867259009 Chr.1: 34868915-34868915 on Build GRCh38
rs751106796 Chr.1: 34868932-34868932 on Build GRCh38
rs142976280 Chr.1: 34868905-34868905 on Build GRCh38
rs375417289 Chr.1: 34868929-34868929 on Build GRCh38
rs200523465 Chr.1: 34868921-34868921 on Build GRCh38
rs747156955 Chr.1: 34868975-34868975 on Build GRCh38

View Full Product Details