Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65552394-65552643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPK
Entrez Gene ID
64105
Gene Name
centromere protein K
Gene Aliases
AF5alpha, CENP-K, FKSG14, P33, Solt
UniGene
Hs.529778
Gene Chromosome Location
Chr.5: 65488919-65563194 on Build GRCh38
Transcripts
NM_001267038.1 NM_022145.4 XM_011543534.2 XM_011543535.2 XM_011543536.2 XM_011543537.2 XM_011543538.2 XM_011543539.2 XM_011543540.2 XM_011543541.2 XM_011543542.2 XM_017009690.1 XM_017009691.1 XM_017009692.1 XM_017009693.1 XM_017009694.1 XM_017009695.1 XM_017009696.1 XM_017009697.1 XM_017009698.1 XM_017009699.1 XM_017009700.1 XM_017009701.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400535 c.126A>T p.L42F Chr.5: 65552535-65552535 on Build GRCh38
1069346 c.153C>T p.T51T Chr.5: 65552508-65552508 on Build GRCh38
4671531 c.114T>C p.C38C Chr.5: 65552547-65552547 on Build GRCh38
1248073 c.115C>G p.Q39E Chr.5: 65552546-65552546 on Build GRCh38
5016087 c.157delT p.S53fs*8 Chr.5: 65552504-65552504 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775330841 Chr.5: 65552531-65552531 on Build GRCh38
rs144481454 Chr.5: 65552525-65552525 on Build GRCh38
rs780355384 Chr.5: 65552476-65552476 on Build GRCh38
rs543487501 Chr.5: 65552548-65552548 on Build GRCh38
rs202188046 Chr.5: 65552572-65552572 on Build GRCh38
rs376533258 Chr.5: 65552586-65552586 on Build GRCh38
rs375328074 Chr.5: 65552560-65552560 on Build GRCh38
rs1043343369 Chr.5: 65552547-65552547 on Build GRCh38
rs1025399187 Chr.5: 65552431-65552431 on Build GRCh38
rs749804748 Chr.5: 65552477-65552477 on Build GRCh38
rs3213939 Chr.5: 65552585-65552585 on Build GRCh38
rs138527872 Chr.5: 65552513-65552513 on Build GRCh38
rs746244389 Chr.5: 65552564-65552565 on Build GRCh38
rs148016416 Chr.5: 65552529-65552529 on Build GRCh38
rs984034409 Chr.5: 65552439-65552439 on Build GRCh38
rs900939286 Chr.5: 65552604-65552604 on Build GRCh38
rs779227890 Chr.5: 65552525-65552527 on Build GRCh38
rs188627320 Chr.5: 65552426-65552426 on Build GRCh38
rs199850162 Chr.5: 65552461-65552461 on Build GRCh38
rs191028228 Chr.5: 65552436-65552436 on Build GRCh38
rs755596314 Chr.5: 65552496-65552496 on Build GRCh38
rs750703310 Chr.5: 65552467-65552467 on Build GRCh38
rs756870907 Chr.5: 65552452-65552452 on Build GRCh38
rs757907403 Chr.5: 65552508-65552508 on Build GRCh38
rs369396438 Chr.5: 65552587-65552587 on Build GRCh38
rs201709067 Chr.5: 65552503-65552503 on Build GRCh38
rs756512541 Chr.5: 65552472-65552472 on Build GRCh38
rs752161299 Chr.5: 65552474-65552474 on Build GRCh38
rs757802386 Chr.5: 65552509-65552512 on Build GRCh38
rs745593902 Chr.5: 65552522-65552522 on Build GRCh38
rs755514250 Chr.5: 65552592-65552592 on Build GRCh38
rs183169838 Chr.5: 65552444-65552444 on Build GRCh38
rs762865136 Chr.5: 65552534-65552534 on Build GRCh38
rs946304876 Chr.5: 65552515-65552515 on Build GRCh38
rs141548635 Chr.5: 65552507-65552507 on Build GRCh38
rs746670416 Chr.5: 65552579-65552580 on Build GRCh38
rs376775830 Chr.5: 65552583-65552583 on Build GRCh38
rs3213938 Chr.5: 65552615-65552615 on Build GRCh38
rs909219818 Chr.5: 65552441-65552441 on Build GRCh38
rs764377925 Chr.5: 65552495-65552495 on Build GRCh38
rs751118163 Chr.5: 65552445-65552445 on Build GRCh38
rs200305420 Chr.5: 65552576-65552576 on Build GRCh38

View Full Product Details