Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61302138-61302371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
UniGene
Hs.290758
Gene Chromosome Location
Chr.11: 61299447-61333212 on Build GRCh38
Transcripts
NM_001923.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439245 c.3138C>T p.S1046S Chr.11: 61302334-61302334 on Build GRCh38
6461927 c.3182T>C p.V1061A Chr.11: 61302290-61302290 on Build GRCh38
122530 c.3201G>C p.K1067N Chr.11: 61302271-61302271 on Build GRCh38
4776290 c.3201G>T p.K1067N Chr.11: 61302271-61302271 on Build GRCh38
6641226 c.3142T>C p.Y1048H Chr.11: 61302330-61302330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747712060 Chr.11: 61302300-61302300 on Build GRCh38
rs779439822 Chr.11: 61302319-61302319 on Build GRCh38
rs750382896 Chr.11: 61302241-61302241 on Build GRCh38
rs28720351 Chr.11: 61302262-61302262 on Build GRCh38
rs560588802 Chr.11: 61302349-61302349 on Build GRCh38
rs889449696 Chr.11: 61302198-61302198 on Build GRCh38
rs112738410 Chr.11: 61302256-61302256 on Build GRCh38
rs772412053 Chr.11: 61302346-61302346 on Build GRCh38
rs760883222 Chr.11: 61302350-61302350 on Build GRCh38
rs780649504 Chr.11: 61302284-61302284 on Build GRCh38
rs769285778 Chr.11: 61302312-61302312 on Build GRCh38
rs779921300 Chr.11: 61302255-61302255 on Build GRCh38
rs372159527 Chr.11: 61302235-61302235 on Build GRCh38
rs200814352 Chr.11: 61302214-61302214 on Build GRCh38
rs758316619 Chr.11: 61302248-61302248 on Build GRCh38
rs138003606 Chr.11: 61302340-61302340 on Build GRCh38
rs575538984 Chr.11: 61302184-61302184 on Build GRCh38
rs150781675 Chr.11: 61302215-61302215 on Build GRCh38
rs182128993 Chr.11: 61302326-61302326 on Build GRCh38
rs750454569 Chr.11: 61302167-61302167 on Build GRCh38
rs774488814 Chr.11: 61302183-61302183 on Build GRCh38
rs115359696 Chr.11: 61302199-61302199 on Build GRCh38
rs561941161 Chr.11: 61302181-61302181 on Build GRCh38
rs146926940 Chr.11: 61302268-61302268 on Build GRCh38

View Full Product Details