Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37590420-37590667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf19
Entrez Gene ID
55286
Gene Name
chromosome 4 open reading frame 19
Gene Aliases
-
UniGene
Hs.107527
Gene Chromosome Location
Chr.4: 37453454-37596146 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
733552 c.587G>A p.G196E Chr.4: 37590642-37590642 on Build GRCh38
7250205 c.532G>T p.D178Y Chr.4: 37590587-37590587 on Build GRCh38
4393658 c.419G>T p.G140V Chr.4: 37590474-37590474 on Build GRCh38
274288 c.463A>G p.T155A Chr.4: 37590518-37590518 on Build GRCh38
222908 c.370G>A p.A124T Chr.4: 37590425-37590425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867130985 Chr.4: 37590577-37590577 on Build GRCh38
rs772068143 Chr.4: 37590509-37590509 on Build GRCh38
rs752124793 Chr.4: 37590554-37590554 on Build GRCh38
rs753035577 Chr.4: 37590540-37590540 on Build GRCh38
rs757295730 Chr.4: 37590556-37590556 on Build GRCh38
rs750491820 Chr.4: 37590578-37590578 on Build GRCh38
rs143777851 Chr.4: 37590472-37590472 on Build GRCh38
rs746140329 Chr.4: 37590479-37590479 on Build GRCh38
rs760251868 Chr.4: 37590513-37590513 on Build GRCh38
rs200921992 Chr.4: 37590500-37590500 on Build GRCh38
rs145307767 Chr.4: 37590627-37590627 on Build GRCh38
rs773297693 Chr.4: 37590511-37590511 on Build GRCh38
rs367617219 Chr.4: 37590631-37590631 on Build GRCh38
rs3822186 Chr.4: 37590628-37590628 on Build GRCh38
rs781386024 Chr.4: 37590476-37590476 on Build GRCh38
rs143923373 Chr.4: 37590590-37590590 on Build GRCh38
rs914049776 Chr.4: 37590451-37590451 on Build GRCh38
rs557428243 Chr.4: 37590504-37590504 on Build GRCh38
rs769425715 Chr.4: 37590616-37590616 on Build GRCh38
rs77826980 Chr.4: 37590614-37590614 on Build GRCh38
rs756320364 Chr.4: 37590579-37590579 on Build GRCh38
rs199818714 Chr.4: 37590564-37590564 on Build GRCh38
rs759287092 Chr.4: 37590533-37590533 on Build GRCh38
rs573345124 Chr.4: 37590591-37590591 on Build GRCh38
rs764824936 Chr.4: 37590534-37590534 on Build GRCh38
rs776161268 Chr.4: 37590611-37590611 on Build GRCh38
rs1014850780 Chr.4: 37590610-37590610 on Build GRCh38
rs2973275 Chr.4: 37590506-37590506 on Build GRCh38
rs746307400 Chr.4: 37590633-37590633 on Build GRCh38
rs377062536 Chr.4: 37590469-37590469 on Build GRCh38
rs753126994 Chr.4: 37590448-37590448 on Build GRCh38
rs763238154 Chr.4: 37590543-37590543 on Build GRCh38
rs757383219 Chr.4: 37590474-37590474 on Build GRCh38
rs746986023 Chr.4: 37590605-37590605 on Build GRCh38
rs776120555 Chr.4: 37590529-37590529 on Build GRCh38
rs764568622 Chr.4: 37590548-37590548 on Build GRCh38
rs938766619 Chr.4: 37590487-37590487 on Build GRCh38
rs149005579 Chr.4: 37590464-37590464 on Build GRCh38
rs770635150 Chr.4: 37590520-37590520 on Build GRCh38
rs114589329 Chr.4: 37590453-37590453 on Build GRCh38
rs780145580 Chr.4: 37590580-37590580 on Build GRCh38
rs765677047 Chr.4: 37590446-37590446 on Build GRCh38

View Full Product Details