Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 989945-990192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT3
Entrez Gene ID
58524
Gene Name
doublesex and mab-3 related transcription factor 3
Gene Aliases
DMRTA3
UniGene
Hs.189174
Gene Chromosome Location
Chr.9: 976968-991732 on Build GRCh38
Transcripts
NM_021240.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6903250 c.42_43insGATT p.L16fs*1 Chr.9: 990039-990040 on Build GRCh38
6876554 c.31T>C p.F11L Chr.9: 990028-990028 on Build GRCh38
6903249 c.455-2_455-1insGATT p.? Chr.9: 990039-990040 on Build GRCh38
2733588 c.128C>T p.S43F Chr.9: 990125-990125 on Build GRCh38
2733580 c.69C>T p.D23D Chr.9: 990066-990066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570625531 Chr.9: 990165-990165 on Build GRCh38
rs749394499 Chr.9: 990055-990055 on Build GRCh38
rs750391764 Chr.9: 990109-990109 on Build GRCh38
rs757070266 Chr.9: 990102-990102 on Build GRCh38
rs771327480 Chr.9: 990151-990151 on Build GRCh38
rs761961638 Chr.9: 989998-989998 on Build GRCh38
rs370973993 Chr.9: 990066-990066 on Build GRCh38
rs778040412 Chr.9: 990137-990137 on Build GRCh38
rs751684817 Chr.9: 990056-990056 on Build GRCh38
rs780040401 Chr.9: 989971-989971 on Build GRCh38
rs768926037 Chr.9: 989982-989982 on Build GRCh38
rs1034240145 Chr.9: 989968-989968 on Build GRCh38
rs370071012 Chr.9: 990154-990154 on Build GRCh38
rs775178489 Chr.9: 990163-990163 on Build GRCh38
rs772443802 Chr.9: 990000-990000 on Build GRCh38
rs752402912 Chr.9: 990031-990031 on Build GRCh38
rs749367206 Chr.9: 989977-989977 on Build GRCh38
rs778998093 Chr.9: 990125-990125 on Build GRCh38
rs748198939 Chr.9: 990130-990130 on Build GRCh38
rs766876143 Chr.9: 990012-990012 on Build GRCh38
rs774553177 Chr.9: 989990-989990 on Build GRCh38
rs748395135 Chr.9: 990067-990067 on Build GRCh38
rs758665068 Chr.9: 990134-990134 on Build GRCh38
rs200133883 Chr.9: 990025-990025 on Build GRCh38
rs781176158 Chr.9: 990054-990054 on Build GRCh38
rs763780736 Chr.9: 990097-990097 on Build GRCh38
rs957741485 Chr.9: 990147-990147 on Build GRCh38
rs376969943 Chr.9: 990074-990074 on Build GRCh38
rs986374173 Chr.9: 990159-990159 on Build GRCh38
rs267602324 Chr.9: 990049-990049 on Build GRCh38
rs759070045 Chr.9: 990023-990023 on Build GRCh38
rs753335535 Chr.9: 990016-990016 on Build GRCh38
rs374084588 Chr.9: 990158-990158 on Build GRCh38
rs777731430 Chr.9: 990045-990045 on Build GRCh38
rs147120737 Chr.9: 990122-990122 on Build GRCh38
rs188630847 Chr.9: 990110-990110 on Build GRCh38
rs758065161 Chr.9: 990041-990041 on Build GRCh38
rs757191358 Chr.9: 990047-990047 on Build GRCh38
rs777186321 Chr.9: 990081-990081 on Build GRCh38
rs764853044 Chr.9: 990086-990086 on Build GRCh38
rs137904899 Chr.9: 990139-990139 on Build GRCh38
rs767152362 Chr.9: 990108-990108 on Build GRCh38
rs756000490 Chr.9: 990119-990119 on Build GRCh38
rs141838962 Chr.9: 990098-990098 on Build GRCh38
rs139310500 Chr.9: 990085-990085 on Build GRCh38
rs762725107 Chr.9: 990093-990093 on Build GRCh38
rs763720874 Chr.9: 990167-990167 on Build GRCh38
rs751327049 Chr.9: 990046-990046 on Build GRCh38
rs10978001 Chr.9: 990076-990076 on Build GRCh38
rs141017949 Chr.9: 990069-990069 on Build GRCh38
rs980226025 Chr.9: 990088-990088 on Build GRCh38
rs773580768 Chr.9: 990003-990003 on Build GRCh38
rs775090232 Chr.9: 990090-990090 on Build GRCh38

View Full Product Details