Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46247047-46247288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDPF1
Entrez Gene ID
150383
Gene Name
cysteine rich DPF motif domain containing 1
Gene Aliases
C22orf40
UniGene
Hs.103110
Gene Chromosome Location
Chr.22: 46244013-46250679 on Build GRCh38
Transcripts
NM_207327.4 XM_011529960.1 XM_011529962.2 XM_011529963.1 XM_011529964.1 XM_011529965.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3063411 c.125A>T p.E42V Chr.22: 46247210-46247210 on Build GRCh38
6018293 c.123G>A p.E41E Chr.22: 46247212-46247212 on Build GRCh38
4478452 c.223C>T p.P75S Chr.22: 46247112-46247112 on Build GRCh38
1035109 c.185C>T p.S62L Chr.22: 46247150-46247150 on Build GRCh38
3371939 c.222C>T p.G74G Chr.22: 46247113-46247113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201903854 Chr.22: 46247235-46247235 on Build GRCh38
rs779147273 Chr.22: 46247267-46247267 on Build GRCh38
rs764718994 Chr.22: 46247222-46247222 on Build GRCh38
rs374135528 Chr.22: 46247179-46247179 on Build GRCh38
rs1011782680 Chr.22: 46247123-46247123 on Build GRCh38
rs376467747 Chr.22: 46247242-46247242 on Build GRCh38
rs757442846 Chr.22: 46247266-46247266 on Build GRCh38
rs778439435 Chr.22: 46247250-46247250 on Build GRCh38
rs751887621 Chr.22: 46247158-46247158 on Build GRCh38
rs769151755 Chr.22: 46247081-46247081 on Build GRCh38
rs749312559 Chr.22: 46247252-46247252 on Build GRCh38
rs763307321 Chr.22: 46247110-46247110 on Build GRCh38
rs756745050 Chr.22: 46247247-46247247 on Build GRCh38
rs774718385 Chr.22: 46247187-46247187 on Build GRCh38
rs200364193 Chr.22: 46247227-46247227 on Build GRCh38
rs778065748 Chr.22: 46247183-46247183 on Build GRCh38
rs1002793055 Chr.22: 46247178-46247178 on Build GRCh38
rs767279891 Chr.22: 46247240-46247244 on Build GRCh38
rs771221630 Chr.22: 46247186-46247186 on Build GRCh38
rs998772126 Chr.22: 46247109-46247109 on Build GRCh38
rs746777787 Chr.22: 46247202-46247202 on Build GRCh38
rs202102175 Chr.22: 46247238-46247238 on Build GRCh38
rs773547172 Chr.22: 46247108-46247108 on Build GRCh38
rs200519607 Chr.22: 46247238-46247239 on Build GRCh38
rs758111260 Chr.22: 46247150-46247150 on Build GRCh38
rs377202462 Chr.22: 46247228-46247228 on Build GRCh38
rs761693028 Chr.22: 46247214-46247214 on Build GRCh38
rs1017646643 Chr.22: 46247123-46247139 on Build GRCh38
rs761337004 Chr.22: 46247131-46247131 on Build GRCh38
rs386822010 Chr.22: 46247235-46247238 on Build GRCh38
rs749511088 Chr.22: 46247185-46247185 on Build GRCh38
rs760399618 Chr.22: 46247118-46247118 on Build GRCh38
rs200244651 Chr.22: 46247236-46247236 on Build GRCh38
rs764772495 Chr.22: 46247143-46247143 on Build GRCh38
rs201564755 Chr.22: 46247237-46247237 on Build GRCh38
rs139000397 Chr.22: 46247155-46247155 on Build GRCh38
rs540181007 Chr.22: 46247149-46247149 on Build GRCh38
rs781692059 Chr.22: 46247169-46247169 on Build GRCh38
rs748655568 Chr.22: 46247171-46247171 on Build GRCh38
rs781558038 Chr.22: 46247096-46247096 on Build GRCh38
rs755351927 Chr.22: 46247239-46247239 on Build GRCh38
rs185615734 Chr.22: 46247070-46247070 on Build GRCh38
rs774276643 Chr.22: 46247234-46247235 on Build GRCh38
rs774507032 Chr.22: 46247111-46247111 on Build GRCh38
rs768358846 Chr.22: 46247204-46247204 on Build GRCh38
rs374811981 Chr.22: 46247176-46247176 on Build GRCh38
rs369413728 Chr.22: 46247243-46247243 on Build GRCh38
rs368537877 Chr.22: 46247235-46247235 on Build GRCh38
rs769790725 Chr.22: 46247097-46247097 on Build GRCh38
rs755332547 Chr.22: 46247166-46247166 on Build GRCh38
rs759612189 Chr.22: 46247217-46247218 on Build GRCh38
rs1026173554 Chr.22: 46247258-46247258 on Build GRCh38
rs769099728 Chr.22: 46247139-46247139 on Build GRCh38
rs55641018 Chr.22: 46247126-46247126 on Build GRCh38
rs764024549 Chr.22: 46247121-46247121 on Build GRCh38
rs369781033 Chr.22: 46247211-46247211 on Build GRCh38

View Full Product Details