Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75028516-75028787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT4
Entrez Gene ID
51301
Gene Name
glucosaminyl (N-acetyl) transferase 4, core 2
Gene Aliases
C2GNT3
UniGene
Hs.272404
Gene Chromosome Location
Chr.5: 75027464-75030899 on Build GRCh38
Transcripts
NM_016591.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
149851 c.1315C>T p.P439S Chr.5: 75028723-75028723 on Build GRCh38
1311313 c.1276G>C p.E426Q Chr.5: 75028762-75028762 on Build GRCh38
1070042 c.1257C>A p.I419I Chr.5: 75028781-75028781 on Build GRCh38
4631043 c.1354A>C p.T452P Chr.5: 75028684-75028684 on Build GRCh38
1176922 c.1260G>T p.L420F Chr.5: 75028778-75028778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025769127 Chr.5: 75028610-75028610 on Build GRCh38
rs749974673 Chr.5: 75028716-75028716 on Build GRCh38
rs35535831 Chr.5: 75028626-75028626 on Build GRCh38
rs779613709 Chr.5: 75028627-75028627 on Build GRCh38
rs755451923 Chr.5: 75028746-75028746 on Build GRCh38
rs755541637 Chr.5: 75028722-75028722 on Build GRCh38
rs926829977 Chr.5: 75028575-75028575 on Build GRCh38
rs763860481 Chr.5: 75028691-75028691 on Build GRCh38
rs762785046 Chr.5: 75028679-75028679 on Build GRCh38
rs780231287 Chr.5: 75028647-75028647 on Build GRCh38
rs751561234 Chr.5: 75028703-75028703 on Build GRCh38
rs189203946 Chr.5: 75028618-75028618 on Build GRCh38
rs779415544 Chr.5: 75028750-75028750 on Build GRCh38
rs760622276 Chr.5: 75028671-75028671 on Build GRCh38
rs3811987 Chr.5: 75028723-75028723 on Build GRCh38
rs767297476 Chr.5: 75028715-75028715 on Build GRCh38
rs542574598 Chr.5: 75028611-75028611 on Build GRCh38
rs771557908 Chr.5: 75028699-75028699 on Build GRCh38
rs772111955 Chr.5: 75028642-75028642 on Build GRCh38
rs1048768155 Chr.5: 75028676-75028676 on Build GRCh38
rs748652005 Chr.5: 75028637-75028637 on Build GRCh38
rs975753879 Chr.5: 75028714-75028714 on Build GRCh38
rs776803653 Chr.5: 75028678-75028678 on Build GRCh38
rs747864988 Chr.5: 75028654-75028654 on Build GRCh38
rs3811986 Chr.5: 75028612-75028612 on Build GRCh38
rs377155784 Chr.5: 75028705-75028705 on Build GRCh38
rs753522564 Chr.5: 75028741-75028741 on Build GRCh38
rs771214880 Chr.5: 75028677-75028677 on Build GRCh38
rs761811317 Chr.5: 75028708-75028708 on Build GRCh38

View Full Product Details