Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45367989-45368214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATM
Entrez Gene ID
2628
Gene Name
glycine amidinotransferase
Gene Aliases
AGAT, AT, CCDS3
UniGene
Hs.560354
Gene Chromosome Location
Chr.15: 45361124-45402317 on Build GRCh38
Transcripts
NM_001321015.1 NM_001482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6022727 c.675+8C>T p.? Chr.15: 45368062-45368062 on Build GRCh38
2189602 c.672C>A p.N224K Chr.15: 45368073-45368073 on Build GRCh38
3501721 c.548C>T p.P183L Chr.15: 45368197-45368197 on Build GRCh38
3501720 c.612C>T p.F204F Chr.15: 45368133-45368133 on Build GRCh38
6077433 c.560G>T p.R187L Chr.15: 45368185-45368185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777156720 Chr.15: 45368140-45368140 on Build GRCh38
rs149853417 Chr.15: 45368121-45368121 on Build GRCh38
rs762437312 Chr.15: 45368055-45368055 on Build GRCh38
rs372760205 Chr.15: 45368024-45368024 on Build GRCh38
rs766041671 Chr.15: 45368061-45368061 on Build GRCh38
rs1049378713 Chr.15: 45368067-45368067 on Build GRCh38
rs953018412 Chr.15: 45368178-45368178 on Build GRCh38
rs755965943 Chr.15: 45368190-45368190 on Build GRCh38
rs191584035 Chr.15: 45368021-45368021 on Build GRCh38
rs397514709 Chr.15: 45368137-45368137 on Build GRCh38
rs748600834 Chr.15: 45368142-45368142 on Build GRCh38
rs759135975 Chr.15: 45368169-45368169 on Build GRCh38
rs778893952 Chr.15: 45368107-45368107 on Build GRCh38
rs764785393 Chr.15: 45368025-45368025 on Build GRCh38
rs181118920 Chr.15: 45368163-45368163 on Build GRCh38
rs112034890 Chr.15: 45368189-45368189 on Build GRCh38
rs374059924 Chr.15: 45368123-45368123 on Build GRCh38
rs752490490 Chr.15: 45368185-45368185 on Build GRCh38
rs773913371 Chr.15: 45368160-45368160 on Build GRCh38
rs751757258 Chr.15: 45368186-45368186 on Build GRCh38
rs377578020 Chr.15: 45368180-45368180 on Build GRCh38
rs752584451 Chr.15: 45368092-45368092 on Build GRCh38
rs752950030 Chr.15: 45368161-45368161 on Build GRCh38
rs565509522 Chr.15: 45368128-45368128 on Build GRCh38
rs151231277 Chr.15: 45368076-45368076 on Build GRCh38
rs769023072 Chr.15: 45368129-45368129 on Build GRCh38
rs377171332 Chr.15: 45368047-45368047 on Build GRCh38
rs759012962 Chr.15: 45368101-45368101 on Build GRCh38
rs796052535 Chr.15: 45368153-45368153 on Build GRCh38
rs774017569 Chr.15: 45368164-45368164 on Build GRCh38
rs111445059 Chr.15: 45368070-45368070 on Build GRCh38
rs780300589 Chr.15: 45368124-45368124 on Build GRCh38
rs770422300 Chr.15: 45368157-45368157 on Build GRCh38
rs772855229 Chr.15: 45368044-45368044 on Build GRCh38
rs369580400 Chr.15: 45368090-45368090 on Build GRCh38
rs903404260 Chr.15: 45368065-45368065 on Build GRCh38

View Full Product Details