Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170986480-170986751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH9
Entrez Gene ID
80133
Gene Name
maestro heat like repeat family member 9
Gene Aliases
ARMC11, C1orf129
UniGene
Hs.591489
Gene Chromosome Location
Chr.1: 170935471-171066341 on Build GRCh38
Transcripts
NM_001163629.1 NM_025063.2 XM_011510005.2 XM_011510006.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478218 c.879G>A p.M293I Chr.1: 170986710-170986710 on Build GRCh38
3478216 c.859C>T p.H287Y Chr.1: 170986690-170986690 on Build GRCh38
3478214 c.838C>T p.L280L Chr.1: 170986669-170986669 on Build GRCh38
7377604 c.781C>T p.L261F Chr.1: 170986612-170986612 on Build GRCh38
2074267 c.817G>A p.A273T Chr.1: 170986648-170986648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751066028 Chr.1: 170986702-170986702 on Build GRCh38
rs772894648 Chr.1: 170986523-170986523 on Build GRCh38
rs976482327 Chr.1: 170986605-170986605 on Build GRCh38
rs756572670 Chr.1: 170986580-170986580 on Build GRCh38
rs759499575 Chr.1: 170986546-170986546 on Build GRCh38
rs769761254 Chr.1: 170986646-170986646 on Build GRCh38
rs775610099 Chr.1: 170986549-170986549 on Build GRCh38
rs143368610 Chr.1: 170986517-170986517 on Build GRCh38
rs186747714 Chr.1: 170986695-170986695 on Build GRCh38
rs767864157 Chr.1: 170986569-170986569 on Build GRCh38
rs941313536 Chr.1: 170986656-170986656 on Build GRCh38
rs963732439 Chr.1: 170986592-170986592 on Build GRCh38
rs764138631 Chr.1: 170986557-170986557 on Build GRCh38
rs972361435 Chr.1: 170986666-170986666 on Build GRCh38
rs767663627 Chr.1: 170986669-170986669 on Build GRCh38
rs757908529 Chr.1: 170986614-170986614 on Build GRCh38
rs778948974 Chr.1: 170986506-170986506 on Build GRCh38
rs774306264 Chr.1: 170986652-170986652 on Build GRCh38
rs756836360 Chr.1: 170986709-170986709 on Build GRCh38
rs762044130 Chr.1: 170986664-170986664 on Build GRCh38
rs745631600 Chr.1: 170986640-170986640 on Build GRCh38
rs780970260 Chr.1: 170986720-170986720 on Build GRCh38
rs749840150 Chr.1: 170986596-170986596 on Build GRCh38
rs768716258 Chr.1: 170986650-170986650 on Build GRCh38
rs745574579 Chr.1: 170986724-170986724 on Build GRCh38
rs941600539 Chr.1: 170986662-170986662 on Build GRCh38
rs780505324 Chr.1: 170986582-170986582 on Build GRCh38
rs3738738 Chr.1: 170986512-170986512 on Build GRCh38
rs200920088 Chr.1: 170986553-170986553 on Build GRCh38
rs552947725 Chr.1: 170986629-170986629 on Build GRCh38
rs982143130 Chr.1: 170986550-170986550 on Build GRCh38
rs372661428 Chr.1: 170986632-170986632 on Build GRCh38
rs765298917 Chr.1: 170986548-170986548 on Build GRCh38
rs903236171 Chr.1: 170986726-170986726 on Build GRCh38
rs530754976 Chr.1: 170986701-170986701 on Build GRCh38
rs755457248 Chr.1: 170986697-170986697 on Build GRCh38
rs757424982 Chr.1: 170986565-170986565 on Build GRCh38
rs769709356 Chr.1: 170986729-170986729 on Build GRCh38
rs369318421 Chr.1: 170986686-170986686 on Build GRCh38
rs754335398 Chr.1: 170986696-170986696 on Build GRCh38
rs369052725 Chr.1: 170986626-170986626 on Build GRCh38
rs541801511 Chr.1: 170986679-170986679 on Build GRCh38
rs751803477 Chr.1: 170986564-170986564 on Build GRCh38
rs754366862 Chr.1: 170986532-170986532 on Build GRCh38
rs564495423 Chr.1: 170986648-170986648 on Build GRCh38
rs565238702 Chr.1: 170986631-170986631 on Build GRCh38
rs750806789 Chr.1: 170986579-170986579 on Build GRCh38
rs922330470 Chr.1: 170986617-170986617 on Build GRCh38
rs1010829633 Chr.1: 170986509-170986509 on Build GRCh38
rs374921240 Chr.1: 170986647-170986647 on Build GRCh38
rs745329125 Chr.1: 170986624-170986625 on Build GRCh38
rs781098754 Chr.1: 170986634-170986634 on Build GRCh38
rs776584776 Chr.1: 170986538-170986538 on Build GRCh38
rs16863907 Chr.1: 170986529-170986529 on Build GRCh38

View Full Product Details