Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67854566-67854825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
UniGene
Hs.444200
Gene Chromosome Location
Chr.17: 67825517-67984378 on Build GRCh38
Transcripts
NM_004459.6 NM_182641.3 XM_005257150.3 XM_005257151.3 XM_005257152.2 XM_005257153.3 XM_005257154.3 XM_005257155.3 XM_005257156.3 XM_005257157.3 XM_005257158.3 XM_005257159.2 XM_005257160.2 XM_005257161.3 XM_011524520.2 XM_011524521.2 XM_011524522.2 XM_011524523.2 XM_011524525.2 XM_011524526.2 XM_017024353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5558087 c.1352A>C p.K451T Chr.17: 67854678-67854678 on Build GRCh38
6234327 c.1292G>T p.C431F Chr.17: 67854618-67854618 on Build GRCh38
1246197 c.1264_1266delGAG p.E423delE Chr.17: 67854590-67854592 on Build GRCh38
6147512 c.1419G>A p.L473L Chr.17: 67854745-67854745 on Build GRCh38
707549 c.1407A>T p.K469N Chr.17: 67854733-67854733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025660131 Chr.17: 67854646-67854646 on Build GRCh38
rs563012201 Chr.17: 67854761-67854761 on Build GRCh38
rs373529800 Chr.17: 67854714-67854714 on Build GRCh38
rs762804280 Chr.17: 67854762-67854762 on Build GRCh38
rs142805142 Chr.17: 67854791-67854791 on Build GRCh38
rs767088881 Chr.17: 67854782-67854782 on Build GRCh38
rs375793981 Chr.17: 67854634-67854634 on Build GRCh38
rs769559184 Chr.17: 67854736-67854736 on Build GRCh38
rs773063236 Chr.17: 67854748-67854748 on Build GRCh38
rs779332775 Chr.17: 67854709-67854709 on Build GRCh38
rs749559945 Chr.17: 67854681-67854681 on Build GRCh38
rs778226418 Chr.17: 67854666-67854666 on Build GRCh38
rs771285295 Chr.17: 67854695-67854695 on Build GRCh38
rs753288973 Chr.17: 67854639-67854639 on Build GRCh38
rs756541780 Chr.17: 67854640-67854640 on Build GRCh38
rs759157712 Chr.17: 67854771-67854771 on Build GRCh38
rs751262818 Chr.17: 67854767-67854767 on Build GRCh38
rs756660271 Chr.17: 67854793-67854793 on Build GRCh38
rs766150640 Chr.17: 67854765-67854765 on Build GRCh38
rs544905785 Chr.17: 67854616-67854616 on Build GRCh38
rs754542236 Chr.17: 67854625-67854625 on Build GRCh38
rs146805567 Chr.17: 67854725-67854725 on Build GRCh38
rs761831796 Chr.17: 67854726-67854726 on Build GRCh38

View Full Product Details