Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132862383-132862616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF9
Entrez Gene ID
2661
Gene Name
growth differentiation factor 9
Gene Aliases
-
UniGene
Hs.25022
Gene Chromosome Location
Chr.5: 132861181-132866884 on Build GRCh38
Transcripts
NM_001288824.2 NM_001288825.2 NM_001288826.2 NM_001288827.2 NM_001288828.2 NM_005260.4 XM_005271957.4 XM_006714585.3 XM_011543308.2 XM_011543309.1 XM_011543310.1 XM_011543311.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7221125 c.447C>T p.T149T Chr.5: 132862507-132862507 on Build GRCh38
6670295 c.500C>A p.S167Y Chr.5: 132862454-132862454 on Build GRCh38
3850094 c.457T>G p.L153V Chr.5: 132862497-132862497 on Build GRCh38
4686733 c.505delT p.S169fs*10 Chr.5: 132862449-132862449 on Build GRCh38
6670299 c.475C>A p.L159M Chr.5: 132862479-132862479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544700965 Chr.5: 132862578-132862578 on Build GRCh38
rs189588476 Chr.5: 132862506-132862506 on Build GRCh38
rs777079606 Chr.5: 132862531-132862531 on Build GRCh38
rs748665532 Chr.5: 132862484-132862484 on Build GRCh38
rs111329919 Chr.5: 132862500-132862500 on Build GRCh38
rs747389045 Chr.5: 132862574-132862574 on Build GRCh38
rs1029833498 Chr.5: 132862412-132862412 on Build GRCh38
rs771076958 Chr.5: 132862577-132862577 on Build GRCh38
rs758835342 Chr.5: 132862564-132862565 on Build GRCh38
rs775474471 Chr.5: 132862511-132862511 on Build GRCh38
rs755369587 Chr.5: 132862472-132862472 on Build GRCh38
rs778065992 Chr.5: 132862553-132862553 on Build GRCh38
rs748662338 Chr.5: 132862551-132862551 on Build GRCh38
rs757000432 Chr.5: 132862443-132862443 on Build GRCh38
rs746135619 Chr.5: 132862591-132862591 on Build GRCh38
rs375662096 Chr.5: 132862436-132862436 on Build GRCh38
rs779467227 Chr.5: 132862473-132862473 on Build GRCh38
rs763023910 Chr.5: 132862512-132862512 on Build GRCh38
rs745394707 Chr.5: 132862461-132862461 on Build GRCh38
rs76349024 Chr.5: 132862518-132862518 on Build GRCh38
rs776726149 Chr.5: 132862424-132862424 on Build GRCh38
rs764029206 Chr.5: 132862437-132862437 on Build GRCh38
rs753031203 Chr.5: 132862565-132862565 on Build GRCh38
rs759344850 Chr.5: 132862427-132862427 on Build GRCh38
rs937225371 Chr.5: 132862519-132862519 on Build GRCh38
rs765235023 Chr.5: 132862431-132862431 on Build GRCh38
rs759665658 Chr.5: 132862509-132862509 on Build GRCh38
rs946753855 Chr.5: 132862545-132862545 on Build GRCh38
rs751223860 Chr.5: 132862441-132862441 on Build GRCh38
rs750106022 Chr.5: 132862528-132862528 on Build GRCh38
rs254285 Chr.5: 132862595-132862595 on Build GRCh38
rs878875329 Chr.5: 132862428-132862428 on Build GRCh38
rs868045630 Chr.5: 132862448-132862448 on Build GRCh38
rs527962952 Chr.5: 132862463-132862463 on Build GRCh38
rs779482176 Chr.5: 132862543-132862543 on Build GRCh38
rs759688531 Chr.5: 132862506-132862507 on Build GRCh38
rs1016988873 Chr.5: 132862478-132862478 on Build GRCh38
rs763939436 Chr.5: 132862516-132862516 on Build GRCh38
rs780798132 Chr.5: 132862458-132862458 on Build GRCh38
rs751421758 Chr.5: 132862517-132862517 on Build GRCh38
rs765586472 Chr.5: 132862507-132862508 on Build GRCh38
rs773609934 Chr.5: 132862490-132862490 on Build GRCh38
rs772407418 Chr.5: 132862485-132862485 on Build GRCh38
rs781473198 Chr.5: 132862579-132862579 on Build GRCh38
rs981642299 Chr.5: 132862515-132862515 on Build GRCh38
rs758936986 Chr.5: 132862552-132862552 on Build GRCh38
rs10491279 Chr.5: 132862408-132862408 on Build GRCh38
rs254286 Chr.5: 132862507-132862507 on Build GRCh38
rs752368679 Chr.5: 132862433-132862433 on Build GRCh38
rs755632730 Chr.5: 132862538-132862538 on Build GRCh38

View Full Product Details