Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80676787-80677042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
UniGene
Hs.147027
Gene Chromosome Location
Chr.X: 80669488-80809736 on Build GRCh38
Transcripts
NM_153252.4 XM_005262113.3 XM_017029384.1

SNP Detail

SNP ID Chromosome Location
rs746056253 Chr.X: 80676963-80676963 on Build GRCh38
rs140852252 Chr.X: 80676918-80676918 on Build GRCh38
rs764461771 Chr.X: 80676836-80676836 on Build GRCh38
rs762018167 Chr.X: 80676839-80676839 on Build GRCh38
rs374537740 Chr.X: 80676908-80676908 on Build GRCh38
rs781010314 Chr.X: 80676960-80676960 on Build GRCh38
rs757937340 Chr.X: 80676921-80676921 on Build GRCh38
rs770696972 Chr.X: 80676946-80676946 on Build GRCh38
rs939185547 Chr.X: 80676945-80676945 on Build GRCh38
rs146425236 Chr.X: 80676888-80676888 on Build GRCh38
rs761235446 Chr.X: 80676943-80676957 on Build GRCh38
rs777503508 Chr.X: 80676926-80676926 on Build GRCh38
rs931613393 Chr.X: 80676900-80676900 on Build GRCh38
rs763128944 Chr.X: 80676812-80676812 on Build GRCh38
rs143090181 Chr.X: 80676940-80676940 on Build GRCh38
rs144015236 Chr.X: 80676838-80676838 on Build GRCh38
rs1055391282 Chr.X: 80676957-80676957 on Build GRCh38
rs200751676 Chr.X: 80676917-80676917 on Build GRCh38
rs769966904 Chr.X: 80676966-80676966 on Build GRCh38
rs768063593 Chr.X: 80676840-80676840 on Build GRCh38

View Full Product Details