Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71616623-71616864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR3
Entrez Gene ID
2833
Gene Name
C-X-C motif chemokine receptor 3
Gene Aliases
CD182, CD183, CKR-L2, CMKAR3, GPR9, IP10-R, Mig-R, MigR
UniGene
Hs.198252
Gene Chromosome Location
Chr.X: 71615913-71618517 on Build GRCh38
Transcripts
NM_001142797.1 NM_001504.1 XM_005262256.3 XM_005262257.3 XM_017029435.1 XM_017029436.1

SNP Detail

SNP ID Chromosome Location
rs199927524 Chr.X: 71616834-71616834 on Build GRCh38
rs751858801 Chr.X: 71616806-71616806 on Build GRCh38
rs771498381 Chr.X: 71616699-71616699 on Build GRCh38
rs899336402 Chr.X: 71616827-71616827 on Build GRCh38
rs756411587 Chr.X: 71616826-71616826 on Build GRCh38
rs757700009 Chr.X: 71616665-71616665 on Build GRCh38
rs763193217 Chr.X: 71616755-71616755 on Build GRCh38
rs979816540 Chr.X: 71616732-71616732 on Build GRCh38
rs776323716 Chr.X: 71616709-71616709 on Build GRCh38
rs770806154 Chr.X: 71616703-71616703 on Build GRCh38
rs780544177 Chr.X: 71616833-71616833 on Build GRCh38
rs765282491 Chr.X: 71616745-71616745 on Build GRCh38
rs753318842 Chr.X: 71616838-71616838 on Build GRCh38
rs149567700 Chr.X: 71616701-71616701 on Build GRCh38
rs377241506 Chr.X: 71616748-71616748 on Build GRCh38
rs925284968 Chr.X: 71616800-71616800 on Build GRCh38
rs746424603 Chr.X: 71616683-71616683 on Build GRCh38
rs777121929 Chr.X: 71616841-71616841 on Build GRCh38
rs766391774 Chr.X: 71616837-71616837 on Build GRCh38
rs759360110 Chr.X: 71616723-71616723 on Build GRCh38
rs750808524 Chr.X: 71616821-71616821 on Build GRCh38
rs747669864 Chr.X: 71616694-71616694 on Build GRCh38
rs940771641 Chr.X: 71616759-71616759 on Build GRCh38
rs867269262 Chr.X: 71616840-71616840 on Build GRCh38
rs368171689 Chr.X: 71616726-71616726 on Build GRCh38
rs953490544 Chr.X: 71616711-71616711 on Build GRCh38
rs763985101 Chr.X: 71616686-71616686 on Build GRCh38
rs111516068 Chr.X: 71616795-71616795 on Build GRCh38
rs753816320 Chr.X: 71616692-71616692 on Build GRCh38
rs757392395 Chr.X: 71616810-71616810 on Build GRCh38
rs754323244 Chr.X: 71616662-71616662 on Build GRCh38
rs2230438 Chr.X: 71616761-71616761 on Build GRCh38
rs746625606 Chr.X: 71616702-71616702 on Build GRCh38
rs755573176 Chr.X: 71616692-71616693 on Build GRCh38
rs921976139 Chr.X: 71616708-71616708 on Build GRCh38
rs370669366 Chr.X: 71616767-71616767 on Build GRCh38
rs867495409 Chr.X: 71616766-71616766 on Build GRCh38
rs767736784 Chr.X: 71616814-71616814 on Build GRCh38

View Full Product Details