Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51450134-51450395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
UniGene
Hs.511386
Gene Chromosome Location
Chr.15: 51447724-51622833 on Build GRCh38
Transcripts
NM_001174116.1 NM_001174117.1 NM_015263.3 XM_005254255.1 XM_005254256.1 XM_017022034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6333176 c.8737C>A p.Q2913K Chr.15: 51450293-51450293 on Build GRCh38
3502069 c.8750C>T p.S2917L Chr.15: 51450280-51450280 on Build GRCh38
6645853 c.8756delG p.G2919fs*48 Chr.15: 51450274-51450274 on Build GRCh38
1323953 c.8811G>A p.T2937T Chr.15: 51450219-51450219 on Build GRCh38
4055507 c.8799G>A p.Q2933Q Chr.15: 51450231-51450231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925327563 Chr.15: 51450332-51450332 on Build GRCh38
rs145628830 Chr.15: 51450333-51450333 on Build GRCh38
rs766459411 Chr.15: 51450336-51450336 on Build GRCh38
rs761137297 Chr.15: 51450158-51450159 on Build GRCh38
rs148728346 Chr.15: 51450161-51450161 on Build GRCh38
rs147864837 Chr.15: 51450219-51450219 on Build GRCh38
rs536376656 Chr.15: 51450228-51450228 on Build GRCh38
rs988248426 Chr.15: 51450362-51450362 on Build GRCh38
rs144627629 Chr.15: 51450186-51450186 on Build GRCh38
rs770446727 Chr.15: 51450270-51450270 on Build GRCh38
rs751224567 Chr.15: 51450174-51450174 on Build GRCh38
rs373660948 Chr.15: 51450169-51450169 on Build GRCh38
rs781495241 Chr.15: 51450340-51450340 on Build GRCh38
rs993163712 Chr.15: 51450214-51450214 on Build GRCh38
rs980785933 Chr.15: 51450331-51450331 on Build GRCh38
rs770426518 Chr.15: 51450261-51450261 on Build GRCh38
rs759293999 Chr.15: 51450183-51450183 on Build GRCh38
rs1033834231 Chr.15: 51450285-51450285 on Build GRCh38
rs199576620 Chr.15: 51450319-51450319 on Build GRCh38
rs760406428 Chr.15: 51450318-51450318 on Build GRCh38
rs762424714 Chr.15: 51450263-51450263 on Build GRCh38
rs538594123 Chr.15: 51450156-51450156 on Build GRCh38
rs773928014 Chr.15: 51450277-51450277 on Build GRCh38
rs777623380 Chr.15: 51450216-51450216 on Build GRCh38
rs142904221 Chr.15: 51450172-51450172 on Build GRCh38
rs751542239 Chr.15: 51450339-51450339 on Build GRCh38
rs368274232 Chr.15: 51450293-51450293 on Build GRCh38
rs752443483 Chr.15: 51450197-51450197 on Build GRCh38
rs139405836 Chr.15: 51450175-51450175 on Build GRCh38
rs756077775 Chr.15: 51450201-51450201 on Build GRCh38
rs756530390 Chr.15: 51450361-51450361 on Build GRCh38
rs753811265 Chr.15: 51450328-51450328 on Build GRCh38
rs775269467 Chr.15: 51450309-51450309 on Build GRCh38
rs767120320 Chr.15: 51450299-51450299 on Build GRCh38
rs748480363 Chr.15: 51450260-51450260 on Build GRCh38
rs769191267 Chr.15: 51450241-51450241 on Build GRCh38
rs374931009 Chr.15: 51450220-51450220 on Build GRCh38
rs918043985 Chr.15: 51450322-51450322 on Build GRCh38
rs201770251 Chr.15: 51450246-51450246 on Build GRCh38

View Full Product Details