Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119157818-119158055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL2
Entrez Gene ID
165257
Gene Name
complement C1q like 2
Gene Aliases
C1QTNF10, CTRP10
UniGene
Hs.433493
Gene Chromosome Location
Chr.2: 119156243-119158895 on Build GRCh38
Transcripts
NM_182528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707737 c.302C>A p.P101H Chr.2: 119157968-119157968 on Build GRCh38
6403231 c.322A>G p.K108E Chr.2: 119157948-119157948 on Build GRCh38
3749590 c.337C>A p.R113R Chr.2: 119157933-119157933 on Build GRCh38
4632270 c.273G>C p.G91G Chr.2: 119157997-119157997 on Build GRCh38
5810013 c.330C>T p.D110D Chr.2: 119157940-119157940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760320943 Chr.2: 119157858-119157858 on Build GRCh38
rs554293273 Chr.2: 119157983-119157983 on Build GRCh38
rs895693195 Chr.2: 119157977-119157977 on Build GRCh38
rs752963277 Chr.2: 119158019-119158019 on Build GRCh38
rs937119741 Chr.2: 119157955-119157955 on Build GRCh38
rs750352100 Chr.2: 119157868-119157868 on Build GRCh38
rs758473715 Chr.2: 119157873-119157873 on Build GRCh38
rs748574253 Chr.2: 119157896-119157896 on Build GRCh38
rs1028665125 Chr.2: 119158001-119158001 on Build GRCh38
rs760279009 Chr.2: 119157926-119157926 on Build GRCh38
rs749745611 Chr.2: 119157905-119157905 on Build GRCh38
rs966222359 Chr.2: 119157950-119157950 on Build GRCh38
rs753631540 Chr.2: 119157862-119157862 on Build GRCh38
rs778492472 Chr.2: 119157902-119157902 on Build GRCh38
rs927120716 Chr.2: 119157938-119157938 on Build GRCh38
rs79015645 Chr.2: 119157856-119157856 on Build GRCh38
rs1009931388 Chr.2: 119157978-119157978 on Build GRCh38
rs755186486 Chr.2: 119157894-119157894 on Build GRCh38
rs768401401 Chr.2: 119157928-119157928 on Build GRCh38
rs530953165 Chr.2: 119157850-119157850 on Build GRCh38
rs762827849 Chr.2: 119157993-119157993 on Build GRCh38
rs766398956 Chr.2: 119157994-119157994 on Build GRCh38
rs1057087559 Chr.2: 119157968-119157968 on Build GRCh38
rs770271281 Chr.2: 119157901-119157901 on Build GRCh38
rs754993161 Chr.2: 119158013-119158013 on Build GRCh38
rs761659381 Chr.2: 119157976-119157976 on Build GRCh38
rs775020011 Chr.2: 119157914-119157914 on Build GRCh38
rs751751172 Chr.2: 119157888-119157888 on Build GRCh38
rs761571469 Chr.2: 119157865-119157865 on Build GRCh38
rs756552955 Chr.2: 119158027-119158027 on Build GRCh38
rs939211120 Chr.2: 119158026-119158026 on Build GRCh38
rs550498525 Chr.2: 119157877-119157877 on Build GRCh38
rs190305077 Chr.2: 119157912-119157912 on Build GRCh38
rs889318406 Chr.2: 119157875-119157875 on Build GRCh38
rs759175451 Chr.2: 119157849-119157849 on Build GRCh38
rs967751202 Chr.2: 119157991-119157991 on Build GRCh38
rs1317848 Chr.2: 119157933-119157933 on Build GRCh38
rs370204561 Chr.2: 119158018-119158018 on Build GRCh38
rs1009102754 Chr.2: 119157878-119157878 on Build GRCh38
rs781238787 Chr.2: 119157895-119157895 on Build GRCh38
rs922210107 Chr.2: 119157965-119157965 on Build GRCh38
rs973131377 Chr.2: 119157874-119157874 on Build GRCh38
rs764940568 Chr.2: 119157979-119157979 on Build GRCh38
rs751699255 Chr.2: 119158003-119158003 on Build GRCh38

View Full Product Details