Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27137619-27137874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
136242 c.641C>T p.A214V Chr.2: 27137689-27137689 on Build GRCh38
4500090 c.557C>G p.S186C Chr.2: 27137773-27137773 on Build GRCh38
137354 c.604C>T p.P202S Chr.2: 27137726-27137726 on Build GRCh38
4500450 c.565C>T p.P189S Chr.2: 27137765-27137765 on Build GRCh38
4553792 c.601G>A p.D201N Chr.2: 27137729-27137729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780539367 Chr.2: 27137645-27137645 on Build GRCh38
rs1050563892 Chr.2: 27137736-27137736 on Build GRCh38
rs553792505 Chr.2: 27137805-27137805 on Build GRCh38
rs776561359 Chr.2: 27137694-27137694 on Build GRCh38
rs769807431 Chr.2: 27137850-27137850 on Build GRCh38
rs757157414 Chr.2: 27137754-27137754 on Build GRCh38
rs754447723 Chr.2: 27137644-27137644 on Build GRCh38
rs755241813 Chr.2: 27137845-27137845 on Build GRCh38
rs577365879 Chr.2: 27137758-27137758 on Build GRCh38
rs1053377871 Chr.2: 27137649-27137649 on Build GRCh38
rs775814488 Chr.2: 27137663-27137663 on Build GRCh38
rs754567211 Chr.2: 27137719-27137719 on Build GRCh38
rs752078499 Chr.2: 27137735-27137735 on Build GRCh38
rs567920330 Chr.2: 27137668-27137668 on Build GRCh38
rs17855664 Chr.2: 27137750-27137750 on Build GRCh38
rs144909666 Chr.2: 27137817-27137817 on Build GRCh38
rs142231081 Chr.2: 27137710-27137710 on Build GRCh38
rs140490572 Chr.2: 27137662-27137662 on Build GRCh38
rs755539510 Chr.2: 27137813-27137813 on Build GRCh38
rs374688258 Chr.2: 27137844-27137844 on Build GRCh38
rs760947115 Chr.2: 27137665-27137665 on Build GRCh38
rs552384043 Chr.2: 27137709-27137709 on Build GRCh38
rs375627013 Chr.2: 27137771-27137771 on Build GRCh38
rs370897786 Chr.2: 27137660-27137660 on Build GRCh38
rs201354948 Chr.2: 27137832-27137832 on Build GRCh38
rs780819617 Chr.2: 27137729-27137729 on Build GRCh38
rs765597175 Chr.2: 27137702-27137702 on Build GRCh38
rs769612295 Chr.2: 27137657-27137657 on Build GRCh38
rs773673413 Chr.2: 27137777-27137777 on Build GRCh38
rs200707097 Chr.2: 27137829-27137829 on Build GRCh38
rs3739097 Chr.2: 27137666-27137666 on Build GRCh38
rs774686521 Chr.2: 27137792-27137792 on Build GRCh38
rs766781312 Chr.2: 27137784-27137784 on Build GRCh38
rs527385741 Chr.2: 27137789-27137789 on Build GRCh38
rs759015470 Chr.2: 27137686-27137687 on Build GRCh38
rs756087817 Chr.2: 27137742-27137742 on Build GRCh38
rs137967723 Chr.2: 27137745-27137745 on Build GRCh38
rs749125277 Chr.2: 27137749-27137749 on Build GRCh38
rs745593024 Chr.2: 27137841-27137841 on Build GRCh38
rs767015829 Chr.2: 27137800-27137800 on Build GRCh38
rs781222541 Chr.2: 27137849-27137849 on Build GRCh38
rs773932451 Chr.2: 27137684-27137695 on Build GRCh38
rs770927319 Chr.2: 27137670-27137670 on Build GRCh38
rs762450544 Chr.2: 27137698-27137698 on Build GRCh38
rs555731696 Chr.2: 27137711-27137711 on Build GRCh38
rs201002698 Chr.2: 27137797-27137797 on Build GRCh38
rs1016912134 Chr.2: 27137760-27137760 on Build GRCh38
rs566953660 Chr.2: 27137658-27137658 on Build GRCh38
rs748172179 Chr.2: 27137651-27137651 on Build GRCh38
rs768757773 Chr.2: 27137769-27137769 on Build GRCh38
rs140239397 Chr.2: 27137714-27137714 on Build GRCh38
rs915073303 Chr.2: 27137722-27137722 on Build GRCh38
rs763423591 Chr.2: 27137779-27137779 on Build GRCh38
rs751256748 Chr.2: 27137718-27137718 on Build GRCh38
rs753873885 Chr.2: 27137822-27137822 on Build GRCh38
rs773256177 Chr.2: 27137853-27137853 on Build GRCh38
rs770356521 Chr.2: 27137772-27137772 on Build GRCh38
rs146960355 Chr.2: 27137668-27137669 on Build GRCh38
rs778920111 Chr.2: 27137833-27137833 on Build GRCh38
rs373438615 Chr.2: 27137842-27137842 on Build GRCh38
rs372566599 Chr.2: 27137759-27137759 on Build GRCh38
rs923964923 Chr.2: 27137679-27137679 on Build GRCh38

View Full Product Details