Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15838021-15838264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD38
Entrez Gene ID
952
Gene Name
CD38 molecule
Gene Aliases
ADPRC 1, ADPRC1
UniGene
Hs.479214
Gene Chromosome Location
Chr.4: 15778265-15853243 on Build GRCh38
Transcripts
NM_001775.3 NR_132660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4926821 c.631T>C p.S211P Chr.4: 15838137-15838137 on Build GRCh38
150895 c.638_648del11 p.S213fs*13 Chr.4: 15838144-15838154 on Build GRCh38
1726165 c.586-2A>G p.? Chr.4: 15838090-15838090 on Build GRCh38
6310595 c.586-4T>A p.? Chr.4: 15838088-15838088 on Build GRCh38
3334451 c.645C>A p.I215I Chr.4: 15838151-15838151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747810765 Chr.4: 15838165-15838165 on Build GRCh38
rs758635228 Chr.4: 15838101-15838101 on Build GRCh38
rs761841027 Chr.4: 15838217-15838217 on Build GRCh38
rs746472953 Chr.4: 15838182-15838182 on Build GRCh38
rs201824472 Chr.4: 15838119-15838119 on Build GRCh38
rs372839265 Chr.4: 15838143-15838143 on Build GRCh38
rs778637217 Chr.4: 15838162-15838162 on Build GRCh38
rs772898980 Chr.4: 15838131-15838131 on Build GRCh38
rs749275326 Chr.4: 15838123-15838123 on Build GRCh38
rs759216064 Chr.4: 15838078-15838078 on Build GRCh38
rs1027078103 Chr.4: 15838141-15838141 on Build GRCh38
rs113665761 Chr.4: 15838078-15838078 on Build GRCh38
rs751606737 Chr.4: 15838127-15838127 on Build GRCh38
rs770416881 Chr.4: 15838214-15838215 on Build GRCh38
rs760738411 Chr.4: 15838062-15838062 on Build GRCh38
rs773104477 Chr.4: 15838078-15838079 on Build GRCh38
rs771423105 Chr.4: 15838059-15838059 on Build GRCh38
rs766399411 Chr.4: 15838072-15838072 on Build GRCh38
rs191558369 Chr.4: 15838083-15838083 on Build GRCh38
rs963709543 Chr.4: 15838071-15838071 on Build GRCh38
rs776644065 Chr.4: 15838187-15838187 on Build GRCh38
rs375774131 Chr.4: 15838060-15838060 on Build GRCh38
rs772900482 Chr.4: 15838172-15838172 on Build GRCh38
rs751660224 Chr.4: 15838214-15838214 on Build GRCh38
rs76024486 Chr.4: 15838198-15838198 on Build GRCh38
rs950566 Chr.4: 15838170-15838170 on Build GRCh38
rs11574925 Chr.4: 15838057-15838057 on Build GRCh38
rs201554603 Chr.4: 15838185-15838185 on Build GRCh38
rs776594734 Chr.4: 15838075-15838075 on Build GRCh38
rs757808810 Chr.4: 15838130-15838130 on Build GRCh38
rs769509864 Chr.4: 15838077-15838078 on Build GRCh38
rs756363707 Chr.4: 15838161-15838161 on Build GRCh38
rs773831927 Chr.4: 15838051-15838051 on Build GRCh38
rs994287038 Chr.4: 15838068-15838068 on Build GRCh38
rs752804349 Chr.4: 15838088-15838088 on Build GRCh38
rs11574926 Chr.4: 15838197-15838197 on Build GRCh38
rs775351791 Chr.4: 15838200-15838200 on Build GRCh38
rs746261556 Chr.4: 15838158-15838158 on Build GRCh38
rs763124131 Chr.4: 15838207-15838207 on Build GRCh38

View Full Product Details