Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73957056-73957323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGUOK
Entrez Gene ID
1716
Gene Name
deoxyguanosine kinase
Gene Aliases
MTDPS3, dGK
UniGene
Hs.469022
Gene Chromosome Location
Chr.2: 73926826-73958961 on Build GRCh38
Transcripts
NM_001318859.1 NM_001318860.1 NM_001318861.1 NM_001318862.1 NM_001318863.1 NM_080916.2 NM_080918.2 NR_134893.1 NR_134894.1 NR_134895.1 NR_134896.1 NR_134897.1 NR_134898.1 XM_011532647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059062 c.580C>A p.L194M Chr.2: 73957179-73957179 on Build GRCh38
3840019 c.603G>C p.K201N Chr.2: 73957136-73957136 on Build GRCh38
6272061 c.583G>T p.A195S Chr.2: 73957182-73957182 on Build GRCh38
6266227 c.707+7G>T p.? Chr.2: 73957247-73957247 on Build GRCh38
1409468 c.640A>C p.I214L Chr.2: 73957173-73957173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772174181 Chr.2: 73957194-73957194 on Build GRCh38
rs104893632 Chr.2: 73957212-73957212 on Build GRCh38
rs12621357 Chr.2: 73957091-73957091 on Build GRCh38
rs375774789 Chr.2: 73957237-73957237 on Build GRCh38
rs773216541 Chr.2: 73957288-73957288 on Build GRCh38
rs541642189 Chr.2: 73957188-73957188 on Build GRCh38
rs1039223855 Chr.2: 73957090-73957090 on Build GRCh38
rs753238518 Chr.2: 73957122-73957122 on Build GRCh38
rs371342464 Chr.2: 73957230-73957230 on Build GRCh38
rs746124008 Chr.2: 73957257-73957257 on Build GRCh38
rs932954536 Chr.2: 73957294-73957294 on Build GRCh38
rs886056322 Chr.2: 73957246-73957247 on Build GRCh38
rs116558935 Chr.2: 73957293-73957293 on Build GRCh38
rs970006371 Chr.2: 73957206-73957206 on Build GRCh38
rs866403196 Chr.2: 73957238-73957238 on Build GRCh38
rs756452893 Chr.2: 73957260-73957260 on Build GRCh38
rs749290011 Chr.2: 73957116-73957116 on Build GRCh38
rs577526451 Chr.2: 73957182-73957182 on Build GRCh38
rs544129089 Chr.2: 73957161-73957161 on Build GRCh38
rs1038758699 Chr.2: 73957269-73957269 on Build GRCh38
rs375686551 Chr.2: 73957197-73957197 on Build GRCh38
rs758857130 Chr.2: 73957135-73957135 on Build GRCh38
rs776350723 Chr.2: 73957205-73957205 on Build GRCh38
rs750279491 Chr.2: 73957082-73957082 on Build GRCh38
rs749443990 Chr.2: 73957283-73957283 on Build GRCh38
rs149649618 Chr.2: 73957229-73957229 on Build GRCh38
rs752255665 Chr.2: 73957225-73957225 on Build GRCh38
rs115206553 Chr.2: 73957163-73957163 on Build GRCh38
rs137902450 Chr.2: 73957186-73957186 on Build GRCh38
rs774323790 Chr.2: 73957238-73957239 on Build GRCh38
rs955629513 Chr.2: 73957154-73957154 on Build GRCh38
rs770802920 Chr.2: 73957134-73957135 on Build GRCh38
rs559692031 Chr.2: 73957284-73957284 on Build GRCh38
rs778233697 Chr.2: 73957147-73957147 on Build GRCh38
rs755337079 Chr.2: 73957110-73957110 on Build GRCh38
rs554589234 Chr.2: 73957104-73957104 on Build GRCh38
rs781487451 Chr.2: 73957169-73957169 on Build GRCh38
rs777542320 Chr.2: 73957090-73957091 on Build GRCh38
rs372217661 Chr.2: 73957211-73957211 on Build GRCh38
rs372259679 Chr.2: 73957105-73957105 on Build GRCh38
rs778327766 Chr.2: 73957108-73957108 on Build GRCh38
rs368726172 Chr.2: 73957102-73957102 on Build GRCh38
rs863223948 Chr.2: 73957191-73957191 on Build GRCh38
rs765866127 Chr.2: 73957170-73957170 on Build GRCh38
rs780391440 Chr.2: 73957266-73957266 on Build GRCh38
rs368154530 Chr.2: 73957208-73957208 on Build GRCh38
rs138703428 Chr.2: 73957160-73957160 on Build GRCh38
rs746006471 Chr.2: 73957177-73957177 on Build GRCh38
rs760354653 Chr.2: 73957085-73957085 on Build GRCh38
rs863223949 Chr.2: 73957138-73957139 on Build GRCh38
rs1041975603 Chr.2: 73957180-73957180 on Build GRCh38
rs886037615 Chr.2: 73957142-73957143 on Build GRCh38
rs766030927 Chr.2: 73957202-73957202 on Build GRCh38
rs957798169 Chr.2: 73957196-73957196 on Build GRCh38
rs990170058 Chr.2: 73957136-73957136 on Build GRCh38
rs376820508 Chr.2: 73957226-73957226 on Build GRCh38

View Full Product Details