Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17600513-17600774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTN
Entrez Gene ID
11034
Gene Name
destrin, actin depolymerizing factor
Gene Aliases
ACTDP, ADF, HEL32, bA462D18.2
UniGene
Hs.304192
Gene Chromosome Location
Chr.20: 17569173-17608242 on Build GRCh38
Transcripts
NM_001011546.1 NM_006870.3 XM_011529142.1 XM_011529143.1 XM_011529144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419336 c.16C>T p.Q6* Chr.20: 17600750-17600750 on Build GRCh38
6336811 c.38G>A p.R13H Chr.20: 17600772-17600772 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757691218 Chr.20: 17600703-17600703 on Build GRCh38
rs967945488 Chr.20: 17600610-17600610 on Build GRCh38
rs774619965 Chr.20: 17600726-17600730 on Build GRCh38
rs958959278 Chr.20: 17600646-17600646 on Build GRCh38
rs1018247706 Chr.20: 17600585-17600585 on Build GRCh38
rs1028909189 Chr.20: 17600651-17600651 on Build GRCh38
rs987485274 Chr.20: 17600658-17600658 on Build GRCh38
rs767343745 Chr.20: 17600633-17600633 on Build GRCh38
rs112058455 Chr.20: 17600593-17600593 on Build GRCh38
rs565932189 Chr.20: 17600585-17600587 on Build GRCh38
rs372881083 Chr.20: 17600745-17600745 on Build GRCh38
rs953480421 Chr.20: 17600657-17600657 on Build GRCh38
rs766162318 Chr.20: 17600695-17600695 on Build GRCh38
rs889060504 Chr.20: 17600561-17600561 on Build GRCh38
rs368746357 Chr.20: 17600734-17600734 on Build GRCh38
rs906395594 Chr.20: 17600592-17600592 on Build GRCh38
rs757377777 Chr.20: 17600735-17600735 on Build GRCh38
rs13038541 Chr.20: 17600547-17600547 on Build GRCh38
rs183297224 Chr.20: 17600626-17600626 on Build GRCh38
rs754433720 Chr.20: 17600665-17600665 on Build GRCh38
rs552522895 Chr.20: 17600710-17600710 on Build GRCh38
rs1015296442 Chr.20: 17600602-17600602 on Build GRCh38
rs977614030 Chr.20: 17600728-17600728 on Build GRCh38
rs752871475 Chr.20: 17600704-17600704 on Build GRCh38
rs755884512 Chr.20: 17600694-17600694 on Build GRCh38
rs529038723 Chr.20: 17600539-17600539 on Build GRCh38
rs769061683 Chr.20: 17600704-17600707 on Build GRCh38
rs781776027 Chr.20: 17600738-17600738 on Build GRCh38
rs894900284 Chr.20: 17600534-17600534 on Build GRCh38
rs777846963 Chr.20: 17600733-17600733 on Build GRCh38

View Full Product Details