Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102834122-102834383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
UniGene
Hs.459106
Gene Chromosome Location
Chr.8: 102826302-102864200 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Gene Symbol
LOC107986873
Entrez Gene ID
107986873
Gene Name
forkhead box protein L2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 102810302-102863067 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238928 c.727_728delTT p.F243fs*6 Chr.8: 102834202-102834203 on Build GRCh38
7171394 c.727T>A p.F243I Chr.8: 102834203-102834203 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144533065 Chr.8: 102834246-102834246 on Build GRCh38
rs1047448410 Chr.8: 102834343-102834343 on Build GRCh38
rs1057255978 Chr.8: 102834318-102834318 on Build GRCh38
rs759271031 Chr.8: 102834266-102834266 on Build GRCh38
rs922845733 Chr.8: 102834332-102834332 on Build GRCh38
rs748335532 Chr.8: 102834314-102834314 on Build GRCh38
rs775495978 Chr.8: 102834272-102834272 on Build GRCh38
rs985582991 Chr.8: 102834341-102834341 on Build GRCh38
rs767052295 Chr.8: 102834274-102834274 on Build GRCh38
rs769957250 Chr.8: 102834225-102834225 on Build GRCh38
rs371109138 Chr.8: 102834247-102834247 on Build GRCh38
rs752115513 Chr.8: 102834168-102834168 on Build GRCh38
rs926106611 Chr.8: 102834352-102834352 on Build GRCh38
rs886110595 Chr.8: 102834347-102834347 on Build GRCh38
rs767119599 Chr.8: 102834157-102834157 on Build GRCh38
rs755321663 Chr.8: 102834299-102834299 on Build GRCh38
rs748201398 Chr.8: 102834214-102834214 on Build GRCh38
rs771382286 Chr.8: 102834287-102834287 on Build GRCh38
rs770906926 Chr.8: 102834248-102834248 on Build GRCh38
rs555551526 Chr.8: 102834338-102834338 on Build GRCh38
rs768731284 Chr.8: 102834207-102834207 on Build GRCh38
rs954798528 Chr.8: 102834228-102834228 on Build GRCh38
rs749314557 Chr.8: 102834274-102834274 on Build GRCh38
rs760130008 Chr.8: 102834284-102834284 on Build GRCh38
rs961049507 Chr.8: 102834288-102834288 on Build GRCh38
rs539047104 Chr.8: 102834180-102834180 on Build GRCh38
rs781027608 Chr.8: 102834273-102834274 on Build GRCh38
rs146537666 Chr.8: 102834277-102834277 on Build GRCh38
rs773287598 Chr.8: 102834234-102834234 on Build GRCh38
rs147507945 Chr.8: 102834198-102834198 on Build GRCh38
rs781302698 Chr.8: 102834310-102834310 on Build GRCh38
rs770857189 Chr.8: 102834179-102834181 on Build GRCh38
rs751851094 Chr.8: 102834297-102834297 on Build GRCh38
rs774283137 Chr.8: 102834252-102834252 on Build GRCh38
rs372262719 Chr.8: 102834197-102834197 on Build GRCh38
rs746329083 Chr.8: 102834196-102834196 on Build GRCh38
rs757828781 Chr.8: 102834169-102834169 on Build GRCh38
rs139775878 Chr.8: 102834152-102834152 on Build GRCh38
rs368927109 Chr.8: 102834158-102834158 on Build GRCh38
rs750990187 Chr.8: 102834286-102834286 on Build GRCh38
rs780439920 Chr.8: 102834294-102834294 on Build GRCh38
rs371369934 Chr.8: 102834325-102834325 on Build GRCh38
rs267601687 Chr.8: 102834200-102834200 on Build GRCh38
rs776823846 Chr.8: 102834211-102834211 on Build GRCh38
rs200449495 Chr.8: 102834285-102834285 on Build GRCh38

View Full Product Details