Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25231475-25231736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBB2
Entrez Gene ID
1415
Gene Name
crystallin beta B2
Gene Aliases
CCA2, CRYB2, CRYB2A, CTRCT3, D22S665
UniGene
Hs.373074
Gene Chromosome Location
Chr.22: 25211660-25231869 on Build GRCh38
Transcripts
NM_000496.2 XM_006724141.3 XM_011529900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6224413 c.521G>A p.S174N Chr.22: 25231675-25231675 on Build GRCh38
3405558 c.478C>T p.R160C Chr.22: 25231632-25231632 on Build GRCh38
4002116 c.483G>A p.G161G Chr.22: 25231637-25231637 on Build GRCh38
387283 c.572G>T p.R191L Chr.22: 25231726-25231726 on Build GRCh38
4571591 c.497T>G p.L166R Chr.22: 25231651-25231651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372020060 Chr.22: 25231598-25231598 on Build GRCh38
rs781136219 Chr.22: 25231570-25231570 on Build GRCh38
rs776509242 Chr.22: 25231697-25231697 on Build GRCh38
rs143080187 Chr.22: 25231713-25231713 on Build GRCh38
rs752421861 Chr.22: 25231704-25231704 on Build GRCh38
rs760240932 Chr.22: 25231594-25231594 on Build GRCh38
rs145217335 Chr.22: 25231543-25231543 on Build GRCh38
rs745938679 Chr.22: 25231625-25231625 on Build GRCh38
rs539443672 Chr.22: 25231626-25231626 on Build GRCh38
rs137852880 Chr.22: 25231588-25231588 on Build GRCh38
rs751736346 Chr.22: 25231619-25231619 on Build GRCh38
rs746319664 Chr.22: 25231571-25231571 on Build GRCh38
rs201605132 Chr.22: 25231647-25231647 on Build GRCh38
rs1057518480 Chr.22: 25231714-25231714 on Build GRCh38
rs138071614 Chr.22: 25231661-25231661 on Build GRCh38
rs9624754 Chr.22: 25231536-25231536 on Build GRCh38
rs886041410 Chr.22: 25231717-25231717 on Build GRCh38
rs766269636 Chr.22: 25231603-25231603 on Build GRCh38
rs137852879 Chr.22: 25231585-25231585 on Build GRCh38
rs765024256 Chr.22: 25231702-25231702 on Build GRCh38
rs771002429 Chr.22: 25231567-25231567 on Build GRCh38
rs758280695 Chr.22: 25231706-25231706 on Build GRCh38
rs373339303 Chr.22: 25231579-25231579 on Build GRCh38
rs887265137 Chr.22: 25231601-25231601 on Build GRCh38
rs983613097 Chr.22: 25231507-25231507 on Build GRCh38
rs748599365 Chr.22: 25231654-25231654 on Build GRCh38
rs761406899 Chr.22: 25231586-25231586 on Build GRCh38
rs1014659292 Chr.22: 25231519-25231522 on Build GRCh38
rs753623673 Chr.22: 25231554-25231554 on Build GRCh38
rs778602223 Chr.22: 25231615-25231615 on Build GRCh38
rs780686115 Chr.22: 25231631-25231631 on Build GRCh38
rs749837487 Chr.22: 25231633-25231633 on Build GRCh38
rs765894916 Chr.22: 25231693-25231693 on Build GRCh38
rs781622480 Chr.22: 25231622-25231622 on Build GRCh38
rs916483069 Chr.22: 25231529-25231530 on Build GRCh38
rs754751963 Chr.22: 25231614-25231614 on Build GRCh38
rs772888950 Chr.22: 25231679-25231679 on Build GRCh38
rs757433401 Chr.22: 25231621-25231621 on Build GRCh38
rs1023262728 Chr.22: 25231690-25231690 on Build GRCh38
rs753819177 Chr.22: 25231612-25231612 on Build GRCh38
rs118129164 Chr.22: 25231535-25231535 on Build GRCh38
rs771615312 Chr.22: 25231671-25231671 on Build GRCh38
rs756215806 Chr.22: 25231719-25231719 on Build GRCh38
rs760153063 Chr.22: 25231683-25231683 on Build GRCh38
rs778032252 Chr.22: 25231561-25231561 on Build GRCh38
rs768953807 Chr.22: 25231580-25231580 on Build GRCh38
rs774587276 Chr.22: 25231649-25231649 on Build GRCh38
rs759396008 Chr.22: 25231701-25231701 on Build GRCh38
rs927796825 Chr.22: 25231538-25231541 on Build GRCh38
rs767817964 Chr.22: 25231712-25231712 on Build GRCh38
rs8140949 Chr.22: 25231637-25231637 on Build GRCh38
rs973063850 Chr.22: 25231525-25231525 on Build GRCh38
rs1052519112 Chr.22: 25231547-25231547 on Build GRCh38
rs74315489 Chr.22: 25231617-25231617 on Build GRCh38
rs137852878 Chr.22: 25231572-25231572 on Build GRCh38
rs572524700 Chr.22: 25231505-25231505 on Build GRCh38
rs864309683 Chr.22: 25231710-25231710 on Build GRCh38
rs754805702 Chr.22: 25231558-25231558 on Build GRCh38
rs963011318 Chr.22: 25231522-25231522 on Build GRCh38
rs776207485 Chr.22: 25231573-25231573 on Build GRCh38
rs749917905 Chr.22: 25231592-25231592 on Build GRCh38

View Full Product Details