Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103377930-103378173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTHRC1
Entrez Gene ID
115908
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
UniGene
Hs.405614
Gene Chromosome Location
Chr.8: 103371515-103383005 on Build GRCh38
Transcripts
NM_001256099.1 NM_138455.3 XM_011516824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4587703 c.467G>A p.R156H Chr.8: 103378121-103378121 on Build GRCh38
1202577 c.376T>C p.C126R Chr.8: 103378030-103378030 on Build GRCh38
5408496 c.396G>A p.R132R Chr.8: 103378092-103378092 on Build GRCh38
4428739 c.517G>C p.E173Q Chr.8: 103378171-103378171 on Build GRCh38
5062757 c.404G>A p.C135Y Chr.8: 103378100-103378100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772990788 Chr.8: 103378100-103378100 on Build GRCh38
rs370183649 Chr.8: 103378063-103378063 on Build GRCh38
rs765386910 Chr.8: 103378026-103378026 on Build GRCh38
rs746538268 Chr.8: 103378001-103378001 on Build GRCh38
rs746901634 Chr.8: 103378069-103378069 on Build GRCh38
rs776352520 Chr.8: 103378131-103378132 on Build GRCh38
rs369875487 Chr.8: 103377982-103377982 on Build GRCh38
rs761116465 Chr.8: 103378114-103378114 on Build GRCh38
rs367999587 Chr.8: 103378091-103378091 on Build GRCh38
rs754161448 Chr.8: 103378015-103378015 on Build GRCh38
rs747888820 Chr.8: 103378090-103378090 on Build GRCh38
rs754897420 Chr.8: 103378074-103378074 on Build GRCh38
rs764595813 Chr.8: 103378120-103378120 on Build GRCh38
rs1030298342 Chr.8: 103377979-103377979 on Build GRCh38
rs757475049 Chr.8: 103378018-103378018 on Build GRCh38
rs773861122 Chr.8: 103378111-103378111 on Build GRCh38
rs769625099 Chr.8: 103378095-103378095 on Build GRCh38
rs2687364 Chr.8: 103377996-103377996 on Build GRCh38
rs374472802 Chr.8: 103378078-103378078 on Build GRCh38
rs145062750 Chr.8: 103378121-103378121 on Build GRCh38
rs748845861 Chr.8: 103378102-103378102 on Build GRCh38
rs376856799 Chr.8: 103378002-103378002 on Build GRCh38
rs963678117 Chr.8: 103378049-103378049 on Build GRCh38
rs770323100 Chr.8: 103378109-103378109 on Build GRCh38
rs750559913 Chr.8: 103378048-103378048 on Build GRCh38
rs74728199 Chr.8: 103377966-103377966 on Build GRCh38
rs111649208 Chr.8: 103378138-103378138 on Build GRCh38
rs747332572 Chr.8: 103378013-103378014 on Build GRCh38
rs1025846410 Chr.8: 103377962-103377962 on Build GRCh38
rs371151606 Chr.8: 103377986-103377986 on Build GRCh38
rs141335365 Chr.8: 103378059-103378059 on Build GRCh38
rs966945057 Chr.8: 103378031-103378031 on Build GRCh38

View Full Product Details