Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179378550-179378797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
UniGene
Hs.658505
Gene Chromosome Location
Chr.1: 179365705-179554735 on Build GRCh38
Transcripts
NM_144696.5 NR_073544.1 XM_011509165.1 XM_011509166.2 XM_011509167.2 XM_011509168.1 XM_011509169.1 XM_011509170.1 XM_011509171.1 XM_011509174.1 XM_011509175.1 XM_011509176.1 XM_011509178.2 XM_011509180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4566434 c.375_376GG>AA p.D126N Chr.1: 179378637-179378638 on Build GRCh38
6875801 c.376G>A p.D126N Chr.1: 179378638-179378638 on Build GRCh38
6059916 c.410A>T p.K137I Chr.1: 179378672-179378672 on Build GRCh38
5378645 c.383C>T p.S128F Chr.1: 179378645-179378645 on Build GRCh38
69913 c.464A>T p.Q155L Chr.1: 179378726-179378726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770966341 Chr.1: 179378761-179378761 on Build GRCh38
rs577910959 Chr.1: 179378716-179378716 on Build GRCh38
rs372241740 Chr.1: 179378626-179378626 on Build GRCh38
rs768163079 Chr.1: 179378654-179378654 on Build GRCh38
rs199947499 Chr.1: 179378717-179378717 on Build GRCh38
rs770728862 Chr.1: 179378734-179378751 on Build GRCh38
rs771208639 Chr.1: 179378655-179378655 on Build GRCh38
rs754848254 Chr.1: 179378678-179378678 on Build GRCh38
rs771228361 Chr.1: 179378593-179378593 on Build GRCh38
rs187184596 Chr.1: 179378754-179378754 on Build GRCh38
rs932756953 Chr.1: 179378607-179378607 on Build GRCh38
rs760299077 Chr.1: 179378752-179378752 on Build GRCh38
rs1051587373 Chr.1: 179378609-179378609 on Build GRCh38
rs777848176 Chr.1: 179378699-179378699 on Build GRCh38
rs748932404 Chr.1: 179378639-179378639 on Build GRCh38
rs878869278 Chr.1: 179378580-179378580 on Build GRCh38
rs377083311 Chr.1: 179378694-179378694 on Build GRCh38
rs773225654 Chr.1: 179378666-179378666 on Build GRCh38
rs765367223 Chr.1: 179378693-179378693 on Build GRCh38
rs767139492 Chr.1: 179378664-179378664 on Build GRCh38
rs201696051 Chr.1: 179378750-179378750 on Build GRCh38
rs140658113 Chr.1: 179378667-179378667 on Build GRCh38
rs752014122 Chr.1: 179378705-179378705 on Build GRCh38
rs761222901 Chr.1: 179378656-179378656 on Build GRCh38
rs867269279 Chr.1: 179378734-179378734 on Build GRCh38
rs777002582 Chr.1: 179378617-179378617 on Build GRCh38
rs747522631 Chr.1: 179378591-179378591 on Build GRCh38
rs772875770 Chr.1: 179378751-179378751 on Build GRCh38
rs10798674 Chr.1: 179378697-179378697 on Build GRCh38
rs753543979 Chr.1: 179378676-179378676 on Build GRCh38
rs552545579 Chr.1: 179378715-179378715 on Build GRCh38
rs747850567 Chr.1: 179378721-179378721 on Build GRCh38
rs781695625 Chr.1: 179378713-179378713 on Build GRCh38
rs150327590 Chr.1: 179378710-179378710 on Build GRCh38
rs912739322 Chr.1: 179378638-179378638 on Build GRCh38

View Full Product Details