Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49640389-49640636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT5
Entrez Gene ID
9334
Gene Name
beta-1,4-galactosyltransferase 5
Gene Aliases
B4Gal-T5, BETA4-GALT-IV, beta4Gal-T5, beta4GalT-V, gt-V
UniGene
Hs.370487
Gene Chromosome Location
Chr.20: 49632945-49713884 on Build GRCh38
Transcripts
NM_004776.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212942 c.715C>G p.P239A Chr.20: 49640557-49640557 on Build GRCh38
5365872 c.717G>A p.P239P Chr.20: 49640555-49640555 on Build GRCh38
3707835 c.728G>A p.R243H Chr.20: 49640544-49640544 on Build GRCh38
1734693 c.690G>C p.L230F Chr.20: 49640582-49640582 on Build GRCh38
382230 c.700G>T p.D234Y Chr.20: 49640572-49640572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113757829 Chr.20: 49640599-49640599 on Build GRCh38
rs190757719 Chr.20: 49640483-49640483 on Build GRCh38
rs572673723 Chr.20: 49640465-49640465 on Build GRCh38
rs778699382 Chr.20: 49640537-49640537 on Build GRCh38
rs913161333 Chr.20: 49640472-49640472 on Build GRCh38
rs762990331 Chr.20: 49640485-49640485 on Build GRCh38
rs766946700 Chr.20: 49640534-49640534 on Build GRCh38
rs1042728182 Chr.20: 49640503-49640503 on Build GRCh38
rs766281878 Chr.20: 49640506-49640506 on Build GRCh38
rs143227970 Chr.20: 49640424-49640424 on Build GRCh38
rs529286825 Chr.20: 49640528-49640528 on Build GRCh38
rs748338335 Chr.20: 49640446-49640447 on Build GRCh38
rs947019403 Chr.20: 49640475-49640475 on Build GRCh38
rs752406119 Chr.20: 49640544-49640544 on Build GRCh38
rs751450920 Chr.20: 49640516-49640516 on Build GRCh38
rs421801 Chr.20: 49640612-49640612 on Build GRCh38
rs760754116 Chr.20: 49640441-49640441 on Build GRCh38
rs755706857 Chr.20: 49640545-49640545 on Build GRCh38

View Full Product Details