Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76323351-76323624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC158
Entrez Gene ID
339965
Gene Name
coiled-coil domain containing 158
Gene Aliases
-
UniGene
Hs.529680
Gene Chromosome Location
Chr.4: 76313039-76421920 on Build GRCh38
Transcripts
NM_001042784.1 XM_005262974.4 XM_006714204.3 XM_011531908.2 XM_011531909.2 XM_011531910.2 XM_011531911.1 XM_011531912.2 XM_011531913.1 XM_011531914.2 XM_011531915.2 XM_011531916.2 XM_011531917.1 XM_017008160.1 XM_017008161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3975029 c.3176T>C p.I1059T Chr.4: 76323391-76323391 on Build GRCh38
6621796 c.3171G>A p.P1057P Chr.4: 76323396-76323396 on Build GRCh38
5071430 c.3158A>T p.D1053V Chr.4: 76323409-76323409 on Build GRCh38
1431082 c.3160T>G p.S1054A Chr.4: 76323407-76323407 on Build GRCh38
6571681 c.3194C>T p.T1065I Chr.4: 76323373-76323373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755584180 Chr.4: 76323392-76323392 on Build GRCh38
rs999256598 Chr.4: 76323510-76323510 on Build GRCh38
rs749967645 Chr.4: 76323388-76323388 on Build GRCh38
rs373834843 Chr.4: 76323452-76323452 on Build GRCh38
rs764558257 Chr.4: 76323431-76323431 on Build GRCh38
rs762160483 Chr.4: 76323451-76323451 on Build GRCh38
rs976403603 Chr.4: 76323470-76323470 on Build GRCh38
rs561179500 Chr.4: 76323553-76323553 on Build GRCh38
rs778903354 Chr.4: 76323397-76323397 on Build GRCh38
rs575700475 Chr.4: 76323466-76323466 on Build GRCh38
rs145943208 Chr.4: 76323396-76323396 on Build GRCh38
rs190834560 Chr.4: 76323581-76323581 on Build GRCh38
rs990668502 Chr.4: 76323569-76323569 on Build GRCh38
rs577702974 Chr.4: 76323556-76323556 on Build GRCh38
rs185875802 Chr.4: 76323505-76323505 on Build GRCh38
rs1034721376 Chr.4: 76323521-76323521 on Build GRCh38
rs34664794 Chr.4: 76323454-76323455 on Build GRCh38
rs1046488319 Chr.4: 76323386-76323386 on Build GRCh38
rs540443438 Chr.4: 76323583-76323583 on Build GRCh38
rs201335763 Chr.4: 76323404-76323404 on Build GRCh38
rs750032163 Chr.4: 76323455-76323455 on Build GRCh38
rs1011751774 Chr.4: 76323421-76323421 on Build GRCh38
rs115033808 Chr.4: 76323585-76323585 on Build GRCh38
rs951189023 Chr.4: 76323520-76323520 on Build GRCh38
rs545304292 Chr.4: 76323394-76323394 on Build GRCh38
rs775895856 Chr.4: 76323416-76323416 on Build GRCh38
rs751581242 Chr.4: 76323578-76323579 on Build GRCh38
rs202223106 Chr.4: 76323412-76323412 on Build GRCh38
rs1007184079 Chr.4: 76323415-76323415 on Build GRCh38
rs774751704 Chr.4: 76323441-76323441 on Build GRCh38
rs369076675 Chr.4: 76323417-76323417 on Build GRCh38
rs749198806 Chr.4: 76323395-76323395 on Build GRCh38
rs771346512 Chr.4: 76323408-76323408 on Build GRCh38
rs1018485505 Chr.4: 76323432-76323432 on Build GRCh38
rs375659396 Chr.4: 76323413-76323413 on Build GRCh38
rs1023853815 Chr.4: 76323570-76323570 on Build GRCh38
rs761333782 Chr.4: 76323578-76323578 on Build GRCh38

View Full Product Details