Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70239149-70239374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
UniGene
Hs.590892
Gene Chromosome Location
Chr.6: 70216040-70303083 on Build GRCh38
Transcripts
NM_001851.4 NM_078485.3 XM_011535429.2 XM_011535430.2 XM_017010246.1 XM_017010247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3629343 c.1354G>A p.D452N Chr.6: 70239283-70239283 on Build GRCh38
1487891 c.2087T>A p.I696K Chr.6: 70239279-70239279 on Build GRCh38
4586886 c.1377G>A p.K459K Chr.6: 70239260-70239260 on Build GRCh38
4586885 c.1377G>A p.K459K Chr.6: 70239260-70239260 on Build GRCh38
5678281 c.2102C>A p.P701Q Chr.6: 70239264-70239264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762345040 Chr.6: 70239244-70239244 on Build GRCh38
rs776043758 Chr.6: 70239310-70239311 on Build GRCh38
rs57089808 Chr.6: 70239198-70239198 on Build GRCh38
rs747816190 Chr.6: 70239228-70239228 on Build GRCh38
rs746285315 Chr.6: 70239213-70239213 on Build GRCh38
rs1055153910 Chr.6: 70239269-70239269 on Build GRCh38
rs752472702 Chr.6: 70239295-70239295 on Build GRCh38
rs377300717 Chr.6: 70239214-70239214 on Build GRCh38
rs1028433877 Chr.6: 70239237-70239237 on Build GRCh38
rs746347768 Chr.6: 70239320-70239320 on Build GRCh38
rs369671433 Chr.6: 70239337-70239337 on Build GRCh38
rs776327245 Chr.6: 70239221-70239221 on Build GRCh38
rs758886634 Chr.6: 70239331-70239331 on Build GRCh38
rs115948163 Chr.6: 70239185-70239185 on Build GRCh38
rs779266947 Chr.6: 70239212-70239212 on Build GRCh38
rs1031393703 Chr.6: 70239289-70239289 on Build GRCh38
rs769372116 Chr.6: 70239233-70239233 on Build GRCh38
rs999864737 Chr.6: 70239255-70239255 on Build GRCh38
rs767433607 Chr.6: 70239284-70239284 on Build GRCh38
rs759430122 Chr.6: 70239282-70239282 on Build GRCh38
rs750552442 Chr.6: 70239219-70239219 on Build GRCh38
rs774146890 Chr.6: 70239260-70239260 on Build GRCh38
rs754000677 Chr.6: 70239312-70239312 on Build GRCh38
rs9446213 Chr.6: 70239316-70239316 on Build GRCh38
rs772742013 Chr.6: 70239239-70239239 on Build GRCh38
rs764360670 Chr.6: 70239309-70239309 on Build GRCh38
rs780405337 Chr.6: 70239333-70239333 on Build GRCh38
rs756434503 Chr.6: 70239308-70239308 on Build GRCh38
rs766377239 Chr.6: 70239246-70239246 on Build GRCh38
rs779542601 Chr.6: 70239318-70239318 on Build GRCh38
rs901123016 Chr.6: 70239200-70239200 on Build GRCh38

View Full Product Details