Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152647047-152647272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs782331121 Chr.X: 152647156-152647156 on Build GRCh38
rs782192329 Chr.X: 152647208-152647208 on Build GRCh38
rs782560179 Chr.X: 152647149-152647150 on Build GRCh38
rs781935399 Chr.X: 152647150-152647150 on Build GRCh38
rs898928431 Chr.X: 152647086-152647086 on Build GRCh38
rs781878653 Chr.X: 152647220-152647220 on Build GRCh38
rs781952537 Chr.X: 152647097-152647097 on Build GRCh38
rs782760807 Chr.X: 152647198-152647198 on Build GRCh38
rs374415140 Chr.X: 152647197-152647197 on Build GRCh38
rs782359940 Chr.X: 152647079-152647079 on Build GRCh38
rs201817909 Chr.X: 152647175-152647175 on Build GRCh38
rs782312786 Chr.X: 152647078-152647078 on Build GRCh38
rs782155034 Chr.X: 152647123-152647123 on Build GRCh38
rs782319804 Chr.X: 152647106-152647106 on Build GRCh38
rs782654845 Chr.X: 152647072-152647072 on Build GRCh38
rs782073807 Chr.X: 152647155-152647155 on Build GRCh38
rs782202426 Chr.X: 152647098-152647098 on Build GRCh38
rs782042263 Chr.X: 152647122-152647122 on Build GRCh38
rs993206839 Chr.X: 152647135-152647135 on Build GRCh38
rs782274510 Chr.X: 152647168-152647168 on Build GRCh38
rs781965071 Chr.X: 152647151-152647151 on Build GRCh38
rs887265975 Chr.X: 152647167-152647167 on Build GRCh38

View Full Product Details