Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100160798-100161037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST4
Entrez Gene ID
79690
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
GAL3ST-4
UniGene
Hs.44856
Gene Chromosome Location
Chr.7: 100159242-100168750 on Build GRCh38
Transcripts
NM_024637.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673364 c.588C>A p.Y196* Chr.7: 100160801-100160801 on Build GRCh38
1673365 c.538C>T p.R180C Chr.7: 100160851-100160851 on Build GRCh38
7306665 c.244G>A p.V82I Chr.7: 100160959-100160959 on Build GRCh38
7095434 c.365C>G p.S122C Chr.7: 100160838-100160838 on Build GRCh38
377977 c.531A>T p.S177S Chr.7: 100160858-100160858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987272302 Chr.7: 100160959-100160959 on Build GRCh38
rs771067362 Chr.7: 100160856-100160857 on Build GRCh38
rs776981856 Chr.7: 100160978-100160984 on Build GRCh38
rs763878367 Chr.7: 100160825-100160825 on Build GRCh38
rs150220137 Chr.7: 100160931-100160931 on Build GRCh38
rs550951408 Chr.7: 100160904-100160904 on Build GRCh38
rs761556100 Chr.7: 100160833-100160833 on Build GRCh38
rs528309446 Chr.7: 100160850-100160850 on Build GRCh38
rs764211552 Chr.7: 100160919-100160919 on Build GRCh38
rs201715592 Chr.7: 100160918-100160918 on Build GRCh38
rs770815177 Chr.7: 100160989-100160989 on Build GRCh38
rs751857243 Chr.7: 100160842-100160842 on Build GRCh38
rs970150876 Chr.7: 100160972-100160972 on Build GRCh38
rs561878173 Chr.7: 100160892-100160892 on Build GRCh38
rs767910572 Chr.7: 100160913-100160913 on Build GRCh38
rs745546597 Chr.7: 100160882-100160882 on Build GRCh38
rs375441548 Chr.7: 100161004-100161004 on Build GRCh38
rs765062933 Chr.7: 100160840-100160840 on Build GRCh38
rs759853278 Chr.7: 100160909-100160909 on Build GRCh38
rs776484864 Chr.7: 100160903-100160903 on Build GRCh38
rs374392589 Chr.7: 100160851-100160851 on Build GRCh38
rs767788924 Chr.7: 100160951-100160951 on Build GRCh38
rs780717594 Chr.7: 100160980-100160980 on Build GRCh38
rs754337842 Chr.7: 100160855-100160855 on Build GRCh38
rs779724224 Chr.7: 100160952-100160952 on Build GRCh38
rs775620117 Chr.7: 100161002-100161002 on Build GRCh38
rs773422049 Chr.7: 100160979-100160979 on Build GRCh38
rs769797926 Chr.7: 100160986-100160986 on Build GRCh38
rs759292135 Chr.7: 100161013-100161013 on Build GRCh38
rs369047097 Chr.7: 100160962-100160962 on Build GRCh38
rs151094352 Chr.7: 100160893-100160893 on Build GRCh38
rs757738817 Chr.7: 100160924-100160924 on Build GRCh38
rs1033229310 Chr.7: 100160990-100160990 on Build GRCh38
rs773076380 Chr.7: 100160905-100160905 on Build GRCh38
rs754323863 Chr.7: 100160922-100160922 on Build GRCh38
rs768184712 Chr.7: 100160897-100160897 on Build GRCh38
rs756207900 Chr.7: 100160854-100160854 on Build GRCh38
rs779199112 Chr.7: 100160877-100160877 on Build GRCh38
rs202164744 Chr.7: 100160867-100160867 on Build GRCh38
rs748040239 Chr.7: 100160982-100160982 on Build GRCh38
rs200266180 Chr.7: 100160935-100160935 on Build GRCh38
rs988095356 Chr.7: 100160981-100160982 on Build GRCh38
rs148850474 Chr.7: 100160914-100160914 on Build GRCh38
rs373798097 Chr.7: 100160968-100160968 on Build GRCh38
rs1014454566 Chr.7: 100160955-100160955 on Build GRCh38
rs759792669 Chr.7: 100160849-100160849 on Build GRCh38
rs773271109 Chr.7: 100160988-100160988 on Build GRCh38
rs140969276 Chr.7: 100160894-100160894 on Build GRCh38
rs867327047 Chr.7: 100160821-100160821 on Build GRCh38

View Full Product Details