Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 22582452-22582703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA2
Entrez Gene ID
3170
Gene Name
forkhead box A2
Gene Aliases
HNF3B, TCF3B
UniGene
Hs.155651
Gene Chromosome Location
Chr.20: 22581004-22585463 on Build GRCh38
Transcripts
NM_021784.4 NM_153675.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5698402 c.643G>T p.D215Y Chr.20: 22582581-22582581 on Build GRCh38
5698864 c.737G>A p.G246D Chr.20: 22582487-22582487 on Build GRCh38
5668684 c.760C>A p.R254S Chr.20: 22582464-22582464 on Build GRCh38
6424410 c.585C>A p.F195L Chr.20: 22582639-22582639 on Build GRCh38
295272 c.545T>A p.L182Q Chr.20: 22582679-22582679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746486260 Chr.20: 22582555-22582555 on Build GRCh38
rs777695135 Chr.20: 22582552-22582552 on Build GRCh38
rs774841083 Chr.20: 22582608-22582608 on Build GRCh38
rs756818226 Chr.20: 22582576-22582576 on Build GRCh38
rs918153891 Chr.20: 22582558-22582558 on Build GRCh38
rs200970860 Chr.20: 22582654-22582654 on Build GRCh38
rs766470566 Chr.20: 22582523-22582523 on Build GRCh38
rs751442261 Chr.20: 22582529-22582529 on Build GRCh38
rs772638884 Chr.20: 22582612-22582612 on Build GRCh38
rs957408004 Chr.20: 22582601-22582602 on Build GRCh38
rs376209663 Chr.20: 22582656-22582656 on Build GRCh38
rs768194412 Chr.20: 22582479-22582479 on Build GRCh38
rs940288500 Chr.20: 22582681-22582681 on Build GRCh38
rs773886523 Chr.20: 22582627-22582627 on Build GRCh38
rs752547674 Chr.20: 22582543-22582543 on Build GRCh38
rs372773557 Chr.20: 22582634-22582634 on Build GRCh38
rs760812511 Chr.20: 22582515-22582515 on Build GRCh38
rs201834142 Chr.20: 22582582-22582582 on Build GRCh38
rs745498650 Chr.20: 22582604-22582604 on Build GRCh38
rs758275197 Chr.20: 22582546-22582546 on Build GRCh38
rs1017711939 Chr.20: 22582519-22582519 on Build GRCh38
rs748535239 Chr.20: 22582610-22582610 on Build GRCh38
rs746203062 Chr.20: 22582620-22582621 on Build GRCh38
rs765012987 Chr.20: 22582538-22582538 on Build GRCh38
rs759791855 Chr.20: 22582672-22582672 on Build GRCh38
rs769397287 Chr.20: 22582605-22582605 on Build GRCh38
rs773365412 Chr.20: 22582480-22582480 on Build GRCh38
rs556684561 Chr.20: 22582507-22582507 on Build GRCh38

View Full Product Details