Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 216684-216935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC127
Entrez Gene ID
133957
Gene Name
coiled-coil domain containing 127
Gene Aliases
-
UniGene
Hs.294145
Gene Chromosome Location
Chr.5: 204760-218182 on Build GRCh38
Transcripts
NM_145265.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3827914 c.97C>A p.P33T Chr.5: 216753-216753 on Build GRCh38
7359713 c.36C>G p.I12M Chr.5: 216814-216814 on Build GRCh38
1067017 c.99A>G p.P33P Chr.5: 216751-216751 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750826523 Chr.5: 216741-216741 on Build GRCh38
rs561280577 Chr.5: 216753-216753 on Build GRCh38
rs749443514 Chr.5: 216782-216782 on Build GRCh38
rs755580860 Chr.5: 216847-216847 on Build GRCh38
rs749271031 Chr.5: 216857-216857 on Build GRCh38
rs745592046 Chr.5: 216881-216881 on Build GRCh38
rs753859156 Chr.5: 216729-216729 on Build GRCh38
rs751862099 Chr.5: 216822-216822 on Build GRCh38
rs765782352 Chr.5: 216737-216737 on Build GRCh38
rs765880132 Chr.5: 216802-216802 on Build GRCh38
rs767015598 Chr.5: 216831-216831 on Build GRCh38
rs761918421 Chr.5: 216801-216801 on Build GRCh38
rs770662116 Chr.5: 216785-216785 on Build GRCh38
rs773993400 Chr.5: 216791-216791 on Build GRCh38
rs918081550 Chr.5: 216704-216704 on Build GRCh38
rs749111901 Chr.5: 216780-216780 on Build GRCh38
rs531789668 Chr.5: 216762-216762 on Build GRCh38
rs752252821 Chr.5: 216836-216836 on Build GRCh38
rs769732170 Chr.5: 216765-216765 on Build GRCh38
rs748265377 Chr.5: 216899-216899 on Build GRCh38
rs763534629 Chr.5: 216813-216813 on Build GRCh38
rs757153066 Chr.5: 216861-216861 on Build GRCh38
rs1049702931 Chr.5: 216894-216894 on Build GRCh38
rs754552959 Chr.5: 216760-216760 on Build GRCh38
rs773810827 Chr.5: 216812-216812 on Build GRCh38
rs532996467 Chr.5: 216907-216907 on Build GRCh38
rs747324292 Chr.5: 216796-216796 on Build GRCh38
rs764276999 Chr.5: 216720-216720 on Build GRCh38
rs565353598 Chr.5: 216897-216897 on Build GRCh38
rs779878024 Chr.5: 216751-216751 on Build GRCh38
rs776786754 Chr.5: 216799-216799 on Build GRCh38
rs768983069 Chr.5: 216750-216750 on Build GRCh38
rs889805299 Chr.5: 216903-216903 on Build GRCh38
rs747594573 Chr.5: 216763-216763 on Build GRCh38
rs777709464 Chr.5: 216849-216849 on Build GRCh38
rs766810569 Chr.5: 216814-216814 on Build GRCh38
rs768749474 Chr.5: 216797-216797 on Build GRCh38
rs777621220 Chr.5: 216766-216766 on Build GRCh38
rs543967762 Chr.5: 216863-216863 on Build GRCh38
rs754715275 Chr.5: 216824-216824 on Build GRCh38
rs762441943 Chr.5: 216730-216730 on Build GRCh38
rs111336094 Chr.5: 216823-216823 on Build GRCh38

View Full Product Details