Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21527654-21527891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPX
Entrez Gene ID
80763
Gene Name
spexin hormone
Gene Aliases
C12orf39
UniGene
Hs.130692
Gene Chromosome Location
Chr.12: 21526257-21532947 on Build GRCh38
Transcripts
NM_030572.3 NR_135187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406644 c.146-10G>A p.? Chr.12: 21527717-21527717 on Build GRCh38
937957 c.193G>C p.D65H Chr.12: 21527774-21527774 on Build GRCh38
3688129 c.205C>T p.P69S Chr.12: 21527786-21527786 on Build GRCh38
937956 c.169C>G p.Q57E Chr.12: 21527750-21527750 on Build GRCh38
6041586 c.176G>T p.R59L Chr.12: 21527757-21527757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752515559 Chr.12: 21527803-21527803 on Build GRCh38
rs766800489 Chr.12: 21527788-21527788 on Build GRCh38
rs57439027 Chr.12: 21527705-21527705 on Build GRCh38
rs772766676 Chr.12: 21527777-21527777 on Build GRCh38
rs746260823 Chr.12: 21527761-21527761 on Build GRCh38
rs201972479 Chr.12: 21527759-21527759 on Build GRCh38
rs775544821 Chr.12: 21527853-21527853 on Build GRCh38
rs759405644 Chr.12: 21527794-21527794 on Build GRCh38
rs191557809 Chr.12: 21527806-21527806 on Build GRCh38
rs774663817 Chr.12: 21527791-21527792 on Build GRCh38
rs1007715658 Chr.12: 21527698-21527698 on Build GRCh38
rs556075309 Chr.12: 21527796-21527796 on Build GRCh38
rs993627332 Chr.12: 21527678-21527678 on Build GRCh38
rs752924466 Chr.12: 21527687-21527687 on Build GRCh38
rs561429147 Chr.12: 21527864-21527864 on Build GRCh38
rs371671017 Chr.12: 21527822-21527822 on Build GRCh38
rs756614711 Chr.12: 21527839-21527839 on Build GRCh38
rs776381885 Chr.12: 21527792-21527792 on Build GRCh38
rs941141482 Chr.12: 21527808-21527808 on Build GRCh38
rs763038657 Chr.12: 21527760-21527760 on Build GRCh38
rs541564199 Chr.12: 21527820-21527820 on Build GRCh38
rs933906667 Chr.12: 21527848-21527848 on Build GRCh38
rs867339947 Chr.12: 21527756-21527756 on Build GRCh38
rs781488731 Chr.12: 21527760-21527760 on Build GRCh38
rs770914361 Chr.12: 21527790-21527790 on Build GRCh38
rs760083167 Chr.12: 21527794-21527803 on Build GRCh38
rs1017816596 Chr.12: 21527722-21527722 on Build GRCh38
rs764403407 Chr.12: 21527813-21527813 on Build GRCh38
rs747839085 Chr.12: 21527774-21527774 on Build GRCh38
rs201207493 Chr.12: 21527786-21527786 on Build GRCh38
rs770007860 Chr.12: 21527765-21527765 on Build GRCh38
rs922522572 Chr.12: 21527845-21527845 on Build GRCh38
rs759075097 Chr.12: 21527767-21527767 on Build GRCh38
rs778660034 Chr.12: 21527766-21527766 on Build GRCh38
rs939276820 Chr.12: 21527824-21527824 on Build GRCh38
rs781734941 Chr.12: 21527836-21527836 on Build GRCh38
rs751671889 Chr.12: 21527706-21527706 on Build GRCh38
rs554536527 Chr.12: 21527711-21527711 on Build GRCh38
rs750858688 Chr.12: 21527837-21527837 on Build GRCh38
rs910547891 Chr.12: 21527860-21527860 on Build GRCh38
rs571937074 Chr.12: 21527871-21527871 on Build GRCh38
rs1029730637 Chr.12: 21527781-21527781 on Build GRCh38
rs966713448 Chr.12: 21527749-21527749 on Build GRCh38
rs758659277 Chr.12: 21527690-21527690 on Build GRCh38
rs7966780 Chr.12: 21527675-21527675 on Build GRCh38
rs765516801 Chr.12: 21527679-21527679 on Build GRCh38

View Full Product Details